Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38071070-38071343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP1B1
Entrez Gene ID
1545
Gene Name
cytochrome P450 family 1 subfamily B member 1
Gene Aliases
CP1B, CYPIB1, GLC3A, P4501B1
UniGene
Hs.154654
Gene Chromosome Location
Chr.2: 38067603-38076181 on Build GRCh38
Transcripts
NM_000104.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1306783 c.1099G>T p.D367Y Chr.2: 38071255-38071255 on Build GRCh38
5021125 c.1213G>A p.A405T Chr.2: 38071141-38071141 on Build GRCh38
1249565 c.1102C>T p.R368C Chr.2: 38071252-38071252 on Build GRCh38
3581479 c.1090G>A p.V364M Chr.2: 38071264-38071264 on Build GRCh38
5015775 c.1076A>G p.E359G Chr.2: 38071278-38071278 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775261951 Chr.2: 38071106-38071106 on Build GRCh38
rs869145989 Chr.2: 38071145-38071154 on Build GRCh38
rs775328093 Chr.2: 38071251-38071252 on Build GRCh38
rs72549381 Chr.2: 38071291-38071291 on Build GRCh38
rs764499798 Chr.2: 38071249-38071249 on Build GRCh38
rs55989760 Chr.2: 38071195-38071195 on Build GRCh38
rs761216127 Chr.2: 38071313-38071313 on Build GRCh38
rs768138533 Chr.2: 38071119-38071119 on Build GRCh38
rs745873505 Chr.2: 38071181-38071181 on Build GRCh38
rs771894894 Chr.2: 38071099-38071099 on Build GRCh38
rs985156509 Chr.2: 38071213-38071214 on Build GRCh38
rs760897392 Chr.2: 38071248-38071248 on Build GRCh38
rs587778873 Chr.2: 38071144-38071145 on Build GRCh38
rs745378110 Chr.2: 38071247-38071247 on Build GRCh38
rs750448258 Chr.2: 38071263-38071263 on Build GRCh38
rs781034244 Chr.2: 38071282-38071282 on Build GRCh38
rs200182794 Chr.2: 38071309-38071309 on Build GRCh38
rs776027129 Chr.2: 38071241-38071241 on Build GRCh38
rs940036742 Chr.2: 38071187-38071187 on Build GRCh38
rs141245683 Chr.2: 38071276-38071276 on Build GRCh38
rs757331407 Chr.2: 38071172-38071172 on Build GRCh38
rs775908245 Chr.2: 38071307-38071307 on Build GRCh38
rs370488779 Chr.2: 38071141-38071141 on Build GRCh38
rs772853286 Chr.2: 38071293-38071293 on Build GRCh38
rs916479886 Chr.2: 38071255-38071255 on Build GRCh38
rs4398252 Chr.2: 38071240-38071240 on Build GRCh38
rs104893622 Chr.2: 38071234-38071234 on Build GRCh38
rs56010818 Chr.2: 38071185-38071185 on Build GRCh38
rs72549378 Chr.2: 38071158-38071158 on Build GRCh38
rs369661494 Chr.2: 38071235-38071235 on Build GRCh38
rs778934619 Chr.2: 38071178-38071178 on Build GRCh38
rs148461905 Chr.2: 38071265-38071265 on Build GRCh38
rs71541925 Chr.2: 38071121-38071121 on Build GRCh38
rs776212230 Chr.2: 38071127-38071127 on Build GRCh38
rs1035845063 Chr.2: 38071150-38071150 on Build GRCh38
rs775035231 Chr.2: 38071140-38071140 on Build GRCh38
rs886055997 Chr.2: 38071152-38071152 on Build GRCh38
rs1031608836 Chr.2: 38071100-38071100 on Build GRCh38
rs768047511 Chr.2: 38071156-38071156 on Build GRCh38
rs1003079068 Chr.2: 38071108-38071108 on Build GRCh38
rs1055330213 Chr.2: 38071244-38071244 on Build GRCh38
rs761190485 Chr.2: 38071134-38071134 on Build GRCh38
rs947487251 Chr.2: 38071278-38071278 on Build GRCh38
rs753971512 Chr.2: 38071171-38071171 on Build GRCh38
rs72549379 Chr.2: 38071264-38071264 on Build GRCh38
rs754048211 Chr.2: 38071250-38071250 on Build GRCh38
rs747810555 Chr.2: 38071290-38071290 on Build GRCh38
rs760381965 Chr.2: 38071114-38071114 on Build GRCh38
rs756363215 Chr.2: 38071164-38071164 on Build GRCh38
rs148542782 Chr.2: 38071186-38071186 on Build GRCh38
rs72481806 Chr.2: 38071159-38071159 on Build GRCh38
rs906925939 Chr.2: 38071273-38071273 on Build GRCh38
rs190572437 Chr.2: 38071298-38071298 on Build GRCh38
rs920199421 Chr.2: 38071201-38071201 on Build GRCh38
rs56305281 Chr.2: 38071218-38071218 on Build GRCh38
rs769250429 Chr.2: 38071217-38071217 on Build GRCh38
rs764181460 Chr.2: 38071170-38071170 on Build GRCh38
rs55771538 Chr.2: 38071261-38071261 on Build GRCh38
rs79204362 Chr.2: 38071251-38071251 on Build GRCh38
rs957253424 Chr.2: 38071129-38071129 on Build GRCh38
rs772830856 Chr.2: 38071222-38071222 on Build GRCh38
rs557905773 Chr.2: 38071204-38071204 on Build GRCh38
rs72549380 Chr.2: 38071278-38071290 on Build GRCh38
rs771691649 Chr.2: 38071248-38071249 on Build GRCh38
rs72480442 Chr.2: 38071252-38071252 on Build GRCh38
rs764779087 Chr.2: 38071135-38071135 on Build GRCh38
rs747801707 Chr.2: 38071214-38071214 on Build GRCh38

View Full Product Details