Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 30759159-30759416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC189
Entrez Gene ID
90835
Gene Name
coiled-coil domain containing 189
Gene Aliases
C16orf93
UniGene
Hs.724010
Gene Chromosome Location
Chr.16: 30757226-30762277 on Build GRCh38
Transcripts
NM_001014979.2 XM_011545976.1 XM_011545978.1 XM_017023852.1 XM_017023853.1 XM_017023854.1 XM_017023855.1 XM_017023856.1 XM_017023857.1 XM_017023858.1 XM_017023859.1 XM_017023860.1 XM_017023861.1 XM_017023862.1 NM_000294.2 NM_001172432.1

Gene Symbol
PHKG2
Entrez Gene ID
5261
Gene Name
phosphorylase kinase catalytic subunit gamma 2
Gene Aliases
GSD9C
UniGene
Hs.65735
Gene Chromosome Location
Chr.16: 30748299-30761176 on Build GRCh38
Transcripts
NM_001014979.2 XM_011545976.1 XM_011545978.1 XM_017023852.1 XM_017023853.1 XM_017023854.1 XM_017023855.1 XM_017023856.1 XM_017023857.1 XM_017023858.1 XM_017023859.1 XM_017023860.1 XM_017023861.1 XM_017023862.1 NM_000294.2 NM_001172432.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4981337 c.721G>C p.E241Q Chr.16: 30759184-30759184 on Build GRCh38
5660835 c.597G>A p.L199L Chr.16: 30759197-30759197 on Build GRCh38
1198211 c.642C>A p.Y214* Chr.16: 30759357-30759357 on Build GRCh38
1198212 c.531C>A p.Y177* Chr.16: 30759357-30759357 on Build GRCh38
5660834 c.708G>A p.L236L Chr.16: 30759197-30759197 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141615439 Chr.16: 30759220-30759220 on Build GRCh38
rs910311389 Chr.16: 30759351-30759352 on Build GRCh38
rs775026044 Chr.16: 30759255-30759255 on Build GRCh38
rs367827173 Chr.16: 30759244-30759244 on Build GRCh38
rs765679699 Chr.16: 30759317-30759319 on Build GRCh38
rs374769997 Chr.16: 30759243-30759243 on Build GRCh38
rs768280328 Chr.16: 30759351-30759351 on Build GRCh38
rs778739275 Chr.16: 30759325-30759325 on Build GRCh38
rs150540962 Chr.16: 30759340-30759340 on Build GRCh38
rs556104200 Chr.16: 30759285-30759285 on Build GRCh38
rs886051918 Chr.16: 30759251-30759251 on Build GRCh38
rs145518957 Chr.16: 30759208-30759208 on Build GRCh38
rs755996498 Chr.16: 30759202-30759202 on Build GRCh38
rs369330243 Chr.16: 30759195-30759196 on Build GRCh38
rs1030095153 Chr.16: 30759269-30759269 on Build GRCh38
rs763267334 Chr.16: 30759356-30759357 on Build GRCh38
rs1011331358 Chr.16: 30759336-30759336 on Build GRCh38
rs375995977 Chr.16: 30759262-30759262 on Build GRCh38
rs751151606 Chr.16: 30759384-30759385 on Build GRCh38
rs373154657 Chr.16: 30759309-30759309 on Build GRCh38
rs759543408 Chr.16: 30759179-30759179 on Build GRCh38
rs760943927 Chr.16: 30759384-30759384 on Build GRCh38
rs191838893 Chr.16: 30759281-30759281 on Build GRCh38
rs750953688 Chr.16: 30759334-30759334 on Build GRCh38
rs767712119 Chr.16: 30759374-30759374 on Build GRCh38
rs771368044 Chr.16: 30759363-30759363 on Build GRCh38
rs775682412 Chr.16: 30759379-30759379 on Build GRCh38
rs753763872 Chr.16: 30759310-30759310 on Build GRCh38
rs757236599 Chr.16: 30759312-30759312 on Build GRCh38
rs749778320 Chr.16: 30759357-30759357 on Build GRCh38
rs35293079 Chr.16: 30759338-30759339 on Build GRCh38
rs139508776 Chr.16: 30759389-30759389 on Build GRCh38
rs745682011 Chr.16: 30759334-30759334 on Build GRCh38
rs1002584684 Chr.16: 30759380-30759380 on Build GRCh38
rs535279129 Chr.16: 30759356-30759356 on Build GRCh38
rs147982789 Chr.16: 30759215-30759215 on Build GRCh38
rs773862121 Chr.16: 30759265-30759265 on Build GRCh38
rs551789158 Chr.16: 30759180-30759180 on Build GRCh38
rs752747001 Chr.16: 30759186-30759186 on Build GRCh38
rs770369435 Chr.16: 30759264-30759264 on Build GRCh38
rs200532920 Chr.16: 30759369-30759369 on Build GRCh38
rs374419590 Chr.16: 30759339-30759340 on Build GRCh38
rs759831279 Chr.16: 30759368-30759368 on Build GRCh38
rs752763963 Chr.16: 30759303-30759303 on Build GRCh38
rs775634684 Chr.16: 30759305-30759305 on Build GRCh38
rs767655301 Chr.16: 30759297-30759297 on Build GRCh38
rs756188707 Chr.16: 30759304-30759304 on Build GRCh38
rs778632923 Chr.16: 30759226-30759226 on Build GRCh38
rs774725515 Chr.16: 30759364-30759364 on Build GRCh38
rs763609894 Chr.16: 30759272-30759272 on Build GRCh38
rs759760912 Chr.16: 30759296-30759296 on Build GRCh38
rs761807384 Chr.16: 30759221-30759221 on Build GRCh38
rs770706959 Chr.16: 30759216-30759216 on Build GRCh38
rs762177394 Chr.16: 30759212-30759212 on Build GRCh38
rs567910325 Chr.16: 30759337-30759337 on Build GRCh38
rs766770966 Chr.16: 30759357-30759358 on Build GRCh38

View Full Product Details