Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45075870-45076119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GEMIN7
Entrez Gene ID
79760
Gene Name
gem nuclear organelle associated protein 7
Gene Aliases
SIP3
UniGene
Hs.466919
Gene Chromosome Location
Chr.19: 45075728-45091524 on Build GRCh38
Transcripts
XM_005259263.4 NM_145288.1

Gene Symbol
ZNF296
Entrez Gene ID
162979
Gene Name
zinc finger protein 296
Gene Aliases
ZFP296, ZNF342
UniGene
Hs.192237
Gene Chromosome Location
Chr.19: 45071500-45076430 on Build GRCh38
Transcripts
XM_005259263.4 NM_145288.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3535858 c.272G>A p.W91* Chr.19: 45076102-45076102 on Build GRCh38
5904028 c.299-8C>T p.? Chr.19: 45075870-45075870 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778388877 Chr.19: 45076086-45076086 on Build GRCh38
rs763666403 Chr.19: 45076068-45076068 on Build GRCh38
rs759489608 Chr.19: 45076052-45076052 on Build GRCh38
rs202235786 Chr.19: 45075909-45075909 on Build GRCh38
rs1012102975 Chr.19: 45075982-45075982 on Build GRCh38
rs1048817780 Chr.19: 45075931-45075931 on Build GRCh38
rs767407886 Chr.19: 45076058-45076058 on Build GRCh38
rs78494352 Chr.19: 45075971-45075971 on Build GRCh38
rs375110024 Chr.19: 45076067-45076067 on Build GRCh38
rs537294799 Chr.19: 45075969-45075969 on Build GRCh38
rs753498815 Chr.19: 45076070-45076070 on Build GRCh38
rs764230391 Chr.19: 45076085-45076086 on Build GRCh38
rs776095172 Chr.19: 45075900-45075900 on Build GRCh38
rs539548200 Chr.19: 45076017-45076017 on Build GRCh38
rs776997746 Chr.19: 45075906-45075906 on Build GRCh38
rs906622980 Chr.19: 45075995-45075995 on Build GRCh38
rs930984033 Chr.19: 45075902-45075902 on Build GRCh38
rs773494348 Chr.19: 45076045-45076045 on Build GRCh38
rs758915819 Chr.19: 45076090-45076090 on Build GRCh38
rs780433716 Chr.19: 45075894-45075894 on Build GRCh38
rs780447363 Chr.19: 45076098-45076098 on Build GRCh38
rs200957174 Chr.19: 45076088-45076088 on Build GRCh38
rs922462494 Chr.19: 45075975-45075975 on Build GRCh38
rs747381968 Chr.19: 45075896-45075896 on Build GRCh38
rs1031082019 Chr.19: 45076033-45076033 on Build GRCh38
rs772519742 Chr.19: 45075891-45075891 on Build GRCh38
rs769982201 Chr.19: 45076041-45076041 on Build GRCh38
rs759262096 Chr.19: 45076071-45076071 on Build GRCh38
rs370590970 Chr.19: 45076065-45076065 on Build GRCh38
rs150503754 Chr.19: 45075984-45075984 on Build GRCh38

View Full Product Details