Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 37840951-37841176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNLT3
Entrez Gene ID
6990
Gene Name
dynein light chain Tctex-type 3
Gene Aliases
RP3, TCTE1L, TCTEX1L
UniGene
Hs.446392
Gene Chromosome Location
Chr.X: 37838836-37847636 on Build GRCh38
Transcripts
NM_006520.2

SNP Detail

SNP ID Chromosome Location
rs113772277 Chr.X: 37841147-37841147 on Build GRCh38
rs763241494 Chr.X: 37841100-37841100 on Build GRCh38
rs777015433 Chr.X: 37841080-37841080 on Build GRCh38
rs1014092779 Chr.X: 37841033-37841033 on Build GRCh38
rs141761454 Chr.X: 37841079-37841079 on Build GRCh38
rs752639637 Chr.X: 37841134-37841134 on Build GRCh38
rs377132994 Chr.X: 37841119-37841119 on Build GRCh38
rs761916341 Chr.X: 37841081-37841081 on Build GRCh38
rs747645392 Chr.X: 37841036-37841036 on Build GRCh38
rs755585203 Chr.X: 37841138-37841138 on Build GRCh38
rs766214123 Chr.X: 37841004-37841004 on Build GRCh38
rs754909699 Chr.X: 37841010-37841010 on Build GRCh38
rs765437409 Chr.X: 37841083-37841083 on Build GRCh38
rs748975598 Chr.X: 37841050-37841050 on Build GRCh38
rs768723890 Chr.X: 37841075-37841075 on Build GRCh38
rs750677550 Chr.X: 37840986-37840986 on Build GRCh38
rs747375636 Chr.X: 37841072-37841072 on Build GRCh38
rs773514664 Chr.X: 37841092-37841092 on Build GRCh38
rs960667348 Chr.X: 37841142-37841142 on Build GRCh38
rs78918057 Chr.X: 37841104-37841104 on Build GRCh38
rs772379871 Chr.X: 37841053-37841053 on Build GRCh38
rs755638072 Chr.X: 37841039-37841039 on Build GRCh38
rs781056190 Chr.X: 37841013-37841013 on Build GRCh38
rs754790032 Chr.X: 37841114-37841114 on Build GRCh38
rs777288541 Chr.X: 37841146-37841146 on Build GRCh38
rs766546636 Chr.X: 37841102-37841102 on Build GRCh38
rs138637421 Chr.X: 37841060-37841060 on Build GRCh38
rs765497272 Chr.X: 37840981-37840981 on Build GRCh38
rs372820619 Chr.X: 37841071-37841071 on Build GRCh38
rs758226572 Chr.X: 37840988-37840988 on Build GRCh38
rs891938747 Chr.X: 37841023-37841023 on Build GRCh38
rs1011448704 Chr.X: 37841144-37841144 on Build GRCh38
rs777343911 Chr.X: 37841045-37841045 on Build GRCh38
rs751356086 Chr.X: 37841006-37841006 on Build GRCh38

View Full Product Details