Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40104140-40104391 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMF
Entrez Gene ID
90427
Gene Name
Bcl2 modifying factor
Gene Aliases
-
UniGene
Hs.591104
Gene Chromosome Location
Chr.15: 40087890-40108884 on Build GRCh38
Transcripts
NM_001003940.1 NM_001003942.1 NM_001003943.2 NM_033503.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
324817 c.301G>A p.G101S Chr.15: 40104332-40104332 on Build GRCh38
4969686 c.364delG p.E122fs*51 Chr.15: 40104269-40104269 on Build GRCh38
1372600 c.363delC p.E122fs*51 Chr.15: 40104270-40104270 on Build GRCh38
1372599 c.386C>T p.A129V Chr.15: 40104247-40104247 on Build GRCh38
2268999 c.417C>A p.C139* Chr.15: 40104216-40104216 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779694283 Chr.15: 40104270-40104270 on Build GRCh38
rs774384488 Chr.15: 40104311-40104311 on Build GRCh38
rs777529517 Chr.15: 40104367-40104367 on Build GRCh38
rs368266341 Chr.15: 40104363-40104363 on Build GRCh38
rs751650029 Chr.15: 40104201-40104201 on Build GRCh38
rs761346715 Chr.15: 40104350-40104351 on Build GRCh38
rs200855550 Chr.15: 40104345-40104345 on Build GRCh38
rs758140624 Chr.15: 40104269-40104269 on Build GRCh38
rs750031739 Chr.15: 40104257-40104257 on Build GRCh38
rs752982097 Chr.15: 40104233-40104233 on Build GRCh38
rs559943013 Chr.15: 40104339-40104339 on Build GRCh38
rs534818374 Chr.15: 40104276-40104276 on Build GRCh38
rs768111287 Chr.15: 40104231-40104231 on Build GRCh38
rs769881011 Chr.15: 40104185-40104185 on Build GRCh38
rs146377359 Chr.15: 40104243-40104243 on Build GRCh38
rs754316143 Chr.15: 40104326-40104326 on Build GRCh38
rs767516633 Chr.15: 40104316-40104316 on Build GRCh38
rs375576342 Chr.15: 40104197-40104197 on Build GRCh38
rs759729759 Chr.15: 40104189-40104189 on Build GRCh38
rs772994326 Chr.15: 40104299-40104299 on Build GRCh38
rs367707665 Chr.15: 40104335-40104335 on Build GRCh38
rs77642791 Chr.15: 40104320-40104320 on Build GRCh38
rs764497660 Chr.15: 40104325-40104325 on Build GRCh38
rs776093229 Chr.15: 40104319-40104320 on Build GRCh38
rs914026915 Chr.15: 40104355-40104355 on Build GRCh38
rs771211750 Chr.15: 40104309-40104309 on Build GRCh38
rs764640125 Chr.15: 40104181-40104183 on Build GRCh38
rs748058329 Chr.15: 40104286-40104286 on Build GRCh38
rs780598356 Chr.15: 40104350-40104350 on Build GRCh38
rs760034128 Chr.15: 40104229-40104229 on Build GRCh38
rs746581474 Chr.15: 40104271-40104271 on Build GRCh38
rs767889296 Chr.15: 40104196-40104196 on Build GRCh38
rs756455504 Chr.15: 40104240-40104240 on Build GRCh38
rs761704589 Chr.15: 40104173-40104173 on Build GRCh38
rs370787974 Chr.15: 40104172-40104172 on Build GRCh38
rs936693934 Chr.15: 40104298-40104298 on Build GRCh38
rs771673224 Chr.15: 40104167-40104167 on Build GRCh38
rs940757142 Chr.15: 40104259-40104259 on Build GRCh38
rs746887985 Chr.15: 40104171-40104171 on Build GRCh38
rs748051334 Chr.15: 40104352-40104352 on Build GRCh38
rs373897311 Chr.15: 40104273-40104273 on Build GRCh38
rs766172511 Chr.15: 40104337-40104337 on Build GRCh38
rs769591201 Chr.15: 40104290-40104290 on Build GRCh38
rs761236561 Chr.15: 40104179-40104180 on Build GRCh38
rs778676383 Chr.15: 40104166-40104166 on Build GRCh38
rs761096058 Chr.15: 40104324-40104324 on Build GRCh38
rs749452663 Chr.15: 40104301-40104301 on Build GRCh38
rs779718409 Chr.15: 40104168-40104168 on Build GRCh38

View Full Product Details