Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 6165332-6165587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH3
Entrez Gene ID
9265
Gene Name
cytohesin 3
Gene Aliases
ARNO3, GRP1, PSCD3
UniGene
Hs.487479
Gene Chromosome Location
Chr.7: 6161779-6272611 on Build GRCh38
Transcripts
NM_004227.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4904087 c.700G>A p.E234K Chr.7: 6165562-6165562 on Build GRCh38
1091139 c.972+1G>A p.? Chr.7: 6165544-6165544 on Build GRCh38
6639591 c.709C>T p.R237W Chr.7: 6165553-6165553 on Build GRCh38
6639583 c.1031C>T p.T344I Chr.7: 6165369-6165369 on Build GRCh38
5094935 c.677A>G p.E226G Chr.7: 6165585-6165585 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs969672371 Chr.7: 6165534-6165534 on Build GRCh38
rs199616562 Chr.7: 6165359-6165359 on Build GRCh38
rs775062316 Chr.7: 6165445-6165446 on Build GRCh38
rs762836791 Chr.7: 6165530-6165530 on Build GRCh38
rs886677624 Chr.7: 6165403-6165403 on Build GRCh38
rs771504465 Chr.7: 6165356-6165356 on Build GRCh38
rs769422824 Chr.7: 6165408-6165410 on Build GRCh38
rs1048372234 Chr.7: 6165389-6165389 on Build GRCh38
rs781368499 Chr.7: 6165410-6165410 on Build GRCh38
rs765997383 Chr.7: 6165362-6165362 on Build GRCh38
rs374674584 Chr.7: 6165473-6165473 on Build GRCh38
rs938456652 Chr.7: 6165563-6165563 on Build GRCh38
rs769462274 Chr.7: 6165516-6165516 on Build GRCh38
rs746582486 Chr.7: 6165507-6165507 on Build GRCh38
rs527902425 Chr.7: 6165483-6165483 on Build GRCh38
rs764184600 Chr.7: 6165544-6165544 on Build GRCh38
rs370290642 Chr.7: 6165459-6165459 on Build GRCh38
rs368762520 Chr.7: 6165497-6165497 on Build GRCh38
rs755203995 Chr.7: 6165560-6165560 on Build GRCh38
rs201977150 Chr.7: 6165461-6165461 on Build GRCh38
rs764092562 Chr.7: 6165471-6165471 on Build GRCh38
rs1043982421 Chr.7: 6165433-6165433 on Build GRCh38
rs772611315 Chr.7: 6165500-6165500 on Build GRCh38
rs368688727 Chr.7: 6165487-6165487 on Build GRCh38
rs890550461 Chr.7: 6165472-6165472 on Build GRCh38
rs763029750 Chr.7: 6165463-6165463 on Build GRCh38
rs760150846 Chr.7: 6165457-6165457 on Build GRCh38
rs201518525 Chr.7: 6165519-6165519 on Build GRCh38
rs778252087 Chr.7: 6165506-6165506 on Build GRCh38
rs779481325 Chr.7: 6165449-6165449 on Build GRCh38
rs145466510 Chr.7: 6165545-6165545 on Build GRCh38
rs755403030 Chr.7: 6165495-6165495 on Build GRCh38
rs768847509 Chr.7: 6165447-6165447 on Build GRCh38
rs756444576 Chr.7: 6165415-6165415 on Build GRCh38
rs2301910 Chr.7: 6165368-6165368 on Build GRCh38
rs770292584 Chr.7: 6165508-6165508 on Build GRCh38
rs749717900 Chr.7: 6165443-6165443 on Build GRCh38
rs551111893 Chr.7: 6165369-6165369 on Build GRCh38
rs764477386 Chr.7: 6165458-6165459 on Build GRCh38
rs753942351 Chr.7: 6165479-6165479 on Build GRCh38
rs534225978 Chr.7: 6165462-6165462 on Build GRCh38
rs766331224 Chr.7: 6165557-6165557 on Build GRCh38
rs756248347 Chr.7: 6165474-6165474 on Build GRCh38
rs563074951 Chr.7: 6165406-6165406 on Build GRCh38
rs751888653 Chr.7: 6165514-6165514 on Build GRCh38
rs780335334 Chr.7: 6165477-6165477 on Build GRCh38
rs926051090 Chr.7: 6165438-6165438 on Build GRCh38
rs761402109 Chr.7: 6165353-6165353 on Build GRCh38
rs748491211 Chr.7: 6165499-6165499 on Build GRCh38
rs772883077 Chr.7: 6165358-6165358 on Build GRCh38
rs200810511 Chr.7: 6165509-6165509 on Build GRCh38
rs548913253 Chr.7: 6165460-6165460 on Build GRCh38
rs376620986 Chr.7: 6165453-6165453 on Build GRCh38
rs376130808 Chr.7: 6165454-6165454 on Build GRCh38
rs760840529 Chr.7: 6165553-6165553 on Build GRCh38
rs1037661144 Chr.7: 6165446-6165446 on Build GRCh38
rs761333570 Chr.7: 6165440-6165440 on Build GRCh38
rs762512844 Chr.7: 6165449-6165450 on Build GRCh38
rs537958636 Chr.7: 6165514-6165514 on Build GRCh38
rs754113042 Chr.7: 6165558-6165558 on Build GRCh38

View Full Product Details