Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 70522673-70522914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM20
Entrez Gene ID
8748
Gene Name
ADAM metallopeptidase domain 20
Gene Aliases
-
UniGene
Hs.177984
Gene Chromosome Location
Chr.14: 70520817-70564742 on Build GRCh38
Transcripts
NM_003814.4 XM_005268151.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3497553 c.1996C>T p.P666S Chr.14: 70522912-70522912 on Build GRCh38
4052094 c.2031T>C p.S677S Chr.14: 70522877-70522877 on Build GRCh38
3401444 c.2110T>A p.C704S Chr.14: 70522798-70522798 on Build GRCh38
1477735 c.2125G>T p.A709S Chr.14: 70522783-70522783 on Build GRCh38
4979926 c.2215G>T p.G739* Chr.14: 70522693-70522693 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999998694 Chr.14: 70522734-70522734 on Build GRCh38
rs971030772 Chr.14: 70522851-70522851 on Build GRCh38
rs1003980658 Chr.14: 70522858-70522858 on Build GRCh38
rs767551477 Chr.14: 70522808-70522811 on Build GRCh38
rs752039366 Chr.14: 70522826-70522826 on Build GRCh38
rs367749946 Chr.14: 70522838-70522838 on Build GRCh38
rs141912564 Chr.14: 70522755-70522755 on Build GRCh38
rs551271125 Chr.14: 70522729-70522729 on Build GRCh38
rs779428789 Chr.14: 70522731-70522731 on Build GRCh38
rs201701584 Chr.14: 70522696-70522696 on Build GRCh38
rs768158998 Chr.14: 70522750-70522750 on Build GRCh38
rs368238335 Chr.14: 70522762-70522762 on Build GRCh38
rs1024005029 Chr.14: 70522815-70522815 on Build GRCh38
rs909128038 Chr.14: 70522699-70522699 on Build GRCh38
rs879189813 Chr.14: 70522732-70522732 on Build GRCh38
rs759704071 Chr.14: 70522781-70522781 on Build GRCh38
rs771796408 Chr.14: 70522760-70522760 on Build GRCh38
rs760524175 Chr.14: 70522763-70522763 on Build GRCh38
rs753840736 Chr.14: 70522775-70522775 on Build GRCh38
rs778308611 Chr.14: 70522713-70522713 on Build GRCh38
rs747464417 Chr.14: 70522717-70522717 on Build GRCh38
rs1051023601 Chr.14: 70522843-70522843 on Build GRCh38
rs755218280 Chr.14: 70522789-70522790 on Build GRCh38
rs781741105 Chr.14: 70522850-70522850 on Build GRCh38
rs1012830688 Chr.14: 70522785-70522785 on Build GRCh38
rs113965969 Chr.14: 70522798-70522798 on Build GRCh38
rs761473055 Chr.14: 70522758-70522758 on Build GRCh38
rs944817763 Chr.14: 70522703-70522703 on Build GRCh38
rs765439203 Chr.14: 70522783-70522783 on Build GRCh38
rs766395855 Chr.14: 70522772-70522772 on Build GRCh38
rs748875168 Chr.14: 70522733-70522733 on Build GRCh38
rs940640020 Chr.14: 70522726-70522726 on Build GRCh38
rs757695391 Chr.14: 70522846-70522846 on Build GRCh38
rs903029734 Chr.14: 70522723-70522723 on Build GRCh38
rs778220921 Chr.14: 70522814-70522814 on Build GRCh38
rs758710570 Chr.14: 70522811-70522811 on Build GRCh38
rs758796268 Chr.14: 70522708-70522708 on Build GRCh38

View Full Product Details