Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27222851-27223086 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAD
Entrez Gene ID
790
Gene Name
carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase
Gene Aliases
CDG1Z
UniGene
Hs.377010
Gene Chromosome Location
Chr.2: 27217257-27243792 on Build GRCh38
Transcripts
NM_001306079.1 NM_004341.4 XM_005264555.3 XM_006712101.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6330926 c.712C>T p.R238C Chr.2: 27222940-27222940 on Build GRCh38
6618831 c.696C>A p.V232V Chr.2: 27222924-27222924 on Build GRCh38
4997604 c.680C>T p.A227V Chr.2: 27222908-27222908 on Build GRCh38
1580639 c.742G>A p.V248I Chr.2: 27222970-27222970 on Build GRCh38
7085271 c.807G>A p.M269I Chr.2: 27223035-27223035 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs561378746 Chr.2: 27222912-27222912 on Build GRCh38
rs267599313 Chr.2: 27222924-27222924 on Build GRCh38
rs760486956 Chr.2: 27222999-27222999 on Build GRCh38
rs149058073 Chr.2: 27223013-27223013 on Build GRCh38
rs146747331 Chr.2: 27222925-27222925 on Build GRCh38
rs377020329 Chr.2: 27222977-27222977 on Build GRCh38
rs1031546164 Chr.2: 27222940-27222940 on Build GRCh38
rs761928259 Chr.2: 27223048-27223048 on Build GRCh38
rs989938315 Chr.2: 27222949-27222949 on Build GRCh38
rs549104107 Chr.2: 27222879-27222879 on Build GRCh38
rs267599312 Chr.2: 27222911-27222911 on Build GRCh38
rs143980497 Chr.2: 27222943-27222943 on Build GRCh38
rs542805154 Chr.2: 27222914-27222914 on Build GRCh38
rs766455326 Chr.2: 27223017-27223017 on Build GRCh38
rs914122027 Chr.2: 27223063-27223063 on Build GRCh38
rs751293494 Chr.2: 27223063-27223064 on Build GRCh38
rs773478601 Chr.2: 27223007-27223007 on Build GRCh38
rs376848110 Chr.2: 27222897-27222897 on Build GRCh38
rs374198129 Chr.2: 27222941-27222941 on Build GRCh38
rs775918081 Chr.2: 27223041-27223041 on Build GRCh38
rs772034961 Chr.2: 27222964-27222964 on Build GRCh38
rs200562416 Chr.2: 27223059-27223059 on Build GRCh38
rs201033076 Chr.2: 27222882-27222882 on Build GRCh38
rs767960934 Chr.2: 27223066-27223066 on Build GRCh38
rs755381059 Chr.2: 27222908-27222908 on Build GRCh38
rs752043643 Chr.2: 27223026-27223026 on Build GRCh38
rs765381267 Chr.2: 27223005-27223005 on Build GRCh38
rs770474953 Chr.2: 27222926-27222926 on Build GRCh38
rs766175437 Chr.2: 27223060-27223060 on Build GRCh38

View Full Product Details