Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40462707-40462980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFP69B
Entrez Gene ID
65243
Gene Name
ZFP69 zinc finger protein B
Gene Aliases
ZKSCAN23B, ZNF643, ZSCAN54B
UniGene
Hs.133034
Gene Chromosome Location
Chr.1: 40450068-40466765 on Build GRCh38
Transcripts
NM_023070.2 XM_005271136.1 XM_005271137.2 XM_005271138.3 XM_005271139.3 XM_017002147.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5817707 c.640T>A p.F214I Chr.1: 40462930-40462930 on Build GRCh38
3489569 c.945C>T p.A315A Chr.1: 40462929-40462929 on Build GRCh38
4744538 c.609T>G p.S203R Chr.1: 40462800-40462800 on Build GRCh38
4589646 c.646_647insA p.I218fs*10 Chr.1: 40462837-40462838 on Build GRCh38
4744536 c.816T>G p.S272R Chr.1: 40462800-40462800 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764361071 Chr.1: 40462818-40462818 on Build GRCh38
rs774812261 Chr.1: 40462901-40462901 on Build GRCh38
rs754072556 Chr.1: 40462822-40462822 on Build GRCh38
rs761972282 Chr.1: 40462952-40462952 on Build GRCh38
rs773126037 Chr.1: 40462922-40462922 on Build GRCh38
rs758471306 Chr.1: 40462852-40462852 on Build GRCh38
rs143880922 Chr.1: 40462880-40462880 on Build GRCh38
rs927910998 Chr.1: 40462920-40462920 on Build GRCh38
rs760012840 Chr.1: 40462909-40462909 on Build GRCh38
rs148910082 Chr.1: 40462915-40462915 on Build GRCh38
rs768165759 Chr.1: 40462790-40462790 on Build GRCh38
rs139917327 Chr.1: 40462789-40462789 on Build GRCh38
rs143377630 Chr.1: 40462816-40462816 on Build GRCh38
rs150144613 Chr.1: 40462910-40462910 on Build GRCh38
rs370968880 Chr.1: 40462757-40462757 on Build GRCh38
rs140502577 Chr.1: 40462890-40462890 on Build GRCh38
rs527970050 Chr.1: 40462866-40462866 on Build GRCh38
rs757011618 Chr.1: 40462827-40462827 on Build GRCh38
rs754065879 Chr.1: 40462924-40462924 on Build GRCh38
rs760800372 Chr.1: 40462917-40462917 on Build GRCh38
rs116091241 Chr.1: 40462736-40462736 on Build GRCh38
rs745685335 Chr.1: 40462863-40462863 on Build GRCh38
rs373622720 Chr.1: 40462955-40462955 on Build GRCh38
rs151064151 Chr.1: 40462893-40462893 on Build GRCh38
rs947927350 Chr.1: 40462908-40462908 on Build GRCh38
rs768111486 Chr.1: 40462891-40462891 on Build GRCh38
rs768286935 Chr.1: 40462754-40462754 on Build GRCh38
rs771403576 Chr.1: 40462900-40462900 on Build GRCh38
rs778726084 Chr.1: 40462852-40462852 on Build GRCh38
rs371518930 Chr.1: 40462912-40462912 on Build GRCh38
rs955594442 Chr.1: 40462735-40462735 on Build GRCh38
rs750378184 Chr.1: 40462780-40462783 on Build GRCh38
rs763365431 Chr.1: 40462764-40462764 on Build GRCh38
rs766918354 Chr.1: 40462765-40462765 on Build GRCh38
rs774968060 Chr.1: 40462787-40462787 on Build GRCh38
rs61748795 Chr.1: 40462803-40462803 on Build GRCh38
rs868535806 Chr.1: 40462927-40462927 on Build GRCh38

View Full Product Details