Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 86694968-86695229 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DACH2
Entrez Gene ID
117154
Gene Name
dachshund family transcription factor 2
Gene Aliases
-
UniGene
Hs.86603
Gene Chromosome Location
Chr.X: 86148451-86832602 on Build GRCh38
Transcripts
NM_001139514.1 NM_001139515.1 NM_053281.3 XM_011530846.2 XM_011530847.2 XM_011530848.2 XM_017029254.1 XM_017029255.1 XM_017029256.1

SNP Detail

SNP ID Chromosome Location
rs780360763 Chr.X: 86695029-86695029 on Build GRCh38
rs377500654 Chr.X: 86695049-86695049 on Build GRCh38
rs149971925 Chr.X: 86695041-86695041 on Build GRCh38
rs775064284 Chr.X: 86695090-86695090 on Build GRCh38
rs756427916 Chr.X: 86695139-86695139 on Build GRCh38
rs766301515 Chr.X: 86695135-86695135 on Build GRCh38
rs767422322 Chr.X: 86695122-86695122 on Build GRCh38
rs754076988 Chr.X: 86695137-86695137 on Build GRCh38
rs146550500 Chr.X: 86695082-86695082 on Build GRCh38
rs888172719 Chr.X: 86695050-86695050 on Build GRCh38
rs752773672 Chr.X: 86695178-86695178 on Build GRCh38
rs996994077 Chr.X: 86695039-86695039 on Build GRCh38
rs201008232 Chr.X: 86695032-86695032 on Build GRCh38
rs772501404 Chr.X: 86695066-86695066 on Build GRCh38
rs779025810 Chr.X: 86695154-86695154 on Build GRCh38
rs773784830 Chr.X: 86695102-86695102 on Build GRCh38
rs778186383 Chr.X: 86695193-86695193 on Build GRCh38
rs758935453 Chr.X: 86695183-86695183 on Build GRCh38
rs370724502 Chr.X: 86695075-86695075 on Build GRCh38
rs953050196 Chr.X: 86695060-86695060 on Build GRCh38
rs778092826 Chr.X: 86695070-86695070 on Build GRCh38
rs375345867 Chr.X: 86695092-86695092 on Build GRCh38
rs368244272 Chr.X: 86695114-86695114 on Build GRCh38
rs750395124 Chr.X: 86695128-86695128 on Build GRCh38
rs961407439 Chr.X: 86695188-86695188 on Build GRCh38
rs760493151 Chr.X: 86695132-86695132 on Build GRCh38
rs982182905 Chr.X: 86695198-86695198 on Build GRCh38

View Full Product Details