Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15529668-15529899 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F22
Entrez Gene ID
126410
Gene Name
cytochrome P450 family 4 subfamily F member 22
Gene Aliases
ARCI5, INLNE, LI3
UniGene
Hs.156452
Gene Chromosome Location
Chr.19: 15508487-15552317 on Build GRCh38
Transcripts
NM_173483.3 XM_011527692.2 XM_011527693.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1325439 c.279C>A p.H93Q Chr.19: 15529765-15529765 on Build GRCh38
181613 c.314C>T p.P105L Chr.19: 15529800-15529800 on Build GRCh38
3529716 c.351C>T p.P117P Chr.19: 15529837-15529837 on Build GRCh38
6361439 c.352C>A p.L118I Chr.19: 15529838-15529838 on Build GRCh38
175473 c.239C>T p.A80V Chr.19: 15529725-15529725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs571371741 Chr.19: 15529875-15529875 on Build GRCh38
rs765769509 Chr.19: 15529869-15529869 on Build GRCh38
rs748409630 Chr.19: 15529849-15529849 on Build GRCh38
rs1057518087 Chr.19: 15529699-15529699 on Build GRCh38
rs751327927 Chr.19: 15529772-15529772 on Build GRCh38
rs145640942 Chr.19: 15529801-15529801 on Build GRCh38
rs376599468 Chr.19: 15529804-15529804 on Build GRCh38
rs755919126 Chr.19: 15529715-15529715 on Build GRCh38
rs201148124 Chr.19: 15529836-15529836 on Build GRCh38
rs148922687 Chr.19: 15529770-15529770 on Build GRCh38
rs184627303 Chr.19: 15529873-15529873 on Build GRCh38
rs143835816 Chr.19: 15529771-15529771 on Build GRCh38
rs369811073 Chr.19: 15529728-15529728 on Build GRCh38
rs749972738 Chr.19: 15529800-15529800 on Build GRCh38
rs377611165 Chr.19: 15529757-15529757 on Build GRCh38
rs745835354 Chr.19: 15529695-15529695 on Build GRCh38
rs749026805 Chr.19: 15529808-15529808 on Build GRCh38
rs747140140 Chr.19: 15529868-15529868 on Build GRCh38
rs766929198 Chr.19: 15529759-15529759 on Build GRCh38
rs73512652 Chr.19: 15529858-15529858 on Build GRCh38
rs145733350 Chr.19: 15529726-15529726 on Build GRCh38
rs374714583 Chr.19: 15529761-15529761 on Build GRCh38
rs771273780 Chr.19: 15529872-15529872 on Build GRCh38
rs865925497 Chr.19: 15529787-15529787 on Build GRCh38
rs750057163 Chr.19: 15529730-15529730 on Build GRCh38
rs192866147 Chr.19: 15529724-15529724 on Build GRCh38
rs772169421 Chr.19: 15529857-15529857 on Build GRCh38
rs374513909 Chr.19: 15529729-15529729 on Build GRCh38
rs116743462 Chr.19: 15529725-15529725 on Build GRCh38
rs781188486 Chr.19: 15529691-15529691 on Build GRCh38
rs756800456 Chr.19: 15529784-15529784 on Build GRCh38
rs779100501 Chr.19: 15529844-15529844 on Build GRCh38
rs766712300 Chr.19: 15529706-15529706 on Build GRCh38
rs773736322 Chr.19: 15529739-15529739 on Build GRCh38
rs377436971 Chr.19: 15529687-15529687 on Build GRCh38
rs766987265 Chr.19: 15529793-15529793 on Build GRCh38

View Full Product Details