Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 74384664-74384913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EREG
Entrez Gene ID
2069
Gene Name
epiregulin
Gene Aliases
EPR, ER, Ep
UniGene
Hs.115263
Gene Chromosome Location
Chr.4: 74365143-74388760 on Build GRCh38
Transcripts
NM_001432.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3428651 c.434G>A p.R145K Chr.4: 74384732-74384732 on Build GRCh38
4548881 c.459G>A p.K153K Chr.4: 74384757-74384757 on Build GRCh38
1753834 c.473G>T p.R158I Chr.4: 74384771-74384771 on Build GRCh38
1057501 c.439C>T p.R147* Chr.4: 74384737-74384737 on Build GRCh38
1694557 c.463G>A p.E155K Chr.4: 74384761-74384761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138440902 Chr.4: 74384861-74384861 on Build GRCh38
rs777112891 Chr.4: 74384699-74384699 on Build GRCh38
rs939307945 Chr.4: 74384827-74384827 on Build GRCh38
rs777971461 Chr.4: 74384757-74384757 on Build GRCh38
rs760032872 Chr.4: 74384708-74384708 on Build GRCh38
rs201806482 Chr.4: 74384799-74384799 on Build GRCh38
rs202042457 Chr.4: 74384814-74384814 on Build GRCh38
rs370303353 Chr.4: 74384755-74384755 on Build GRCh38
rs771158792 Chr.4: 74384768-74384768 on Build GRCh38
rs34477425 Chr.4: 74384731-74384731 on Build GRCh38
rs139275751 Chr.4: 74384773-74384773 on Build GRCh38
rs373762804 Chr.4: 74384826-74384826 on Build GRCh38
rs749901172 Chr.4: 74384803-74384803 on Build GRCh38
rs781439268 Chr.4: 74384837-74384837 on Build GRCh38
rs760152514 Chr.4: 74384809-74384809 on Build GRCh38
rs572731794 Chr.4: 74384789-74384789 on Build GRCh38
rs145342792 Chr.4: 74384798-74384798 on Build GRCh38
rs867627418 Chr.4: 74384785-74384785 on Build GRCh38
rs751323778 Chr.4: 74384723-74384723 on Build GRCh38
rs775872702 Chr.4: 74384713-74384713 on Build GRCh38
rs757419853 Chr.4: 74384834-74384834 on Build GRCh38
rs763272962 Chr.4: 74384715-74384715 on Build GRCh38
rs867993621 Chr.4: 74384797-74384797 on Build GRCh38
rs747293062 Chr.4: 74384763-74384763 on Build GRCh38
rs368828647 Chr.4: 74384831-74384831 on Build GRCh38
rs948605050 Chr.4: 74384866-74384866 on Build GRCh38
rs1044207528 Chr.4: 74384879-74384879 on Build GRCh38
rs775897941 Chr.4: 74384775-74384775 on Build GRCh38
rs752496685 Chr.4: 74384830-74384830 on Build GRCh38
rs866675158 Chr.4: 74384761-74384761 on Build GRCh38
rs753646680 Chr.4: 74384744-74384744 on Build GRCh38
rs781184714 Chr.4: 74384769-74384769 on Build GRCh38
rs35275884 Chr.4: 74384738-74384738 on Build GRCh38
rs569866282 Chr.4: 74384882-74384882 on Build GRCh38
rs763804198 Chr.4: 74384717-74384717 on Build GRCh38
rs374059228 Chr.4: 74384786-74384786 on Build GRCh38
rs765691644 Chr.4: 74384709-74384709 on Build GRCh38
rs780596944 Chr.4: 74384859-74384859 on Build GRCh38
rs745925676 Chr.4: 74384846-74384846 on Build GRCh38
rs376236920 Chr.4: 74384737-74384737 on Build GRCh38
rs763367714 Chr.4: 74384779-74384779 on Build GRCh38
rs892135942 Chr.4: 74384753-74384753 on Build GRCh38
rs771068928 Chr.4: 74384694-74384694 on Build GRCh38
rs372115806 Chr.4: 74384813-74384813 on Build GRCh38
rs867376757 Chr.4: 74384853-74384853 on Build GRCh38
rs192335022 Chr.4: 74384848-74384848 on Build GRCh38

View Full Product Details