Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136804757-136804992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC183
Entrez Gene ID
84960
Gene Name
coiled-coil domain containing 183
Gene Aliases
KIAA1984, PARF, bA216L13.7
UniGene
Hs.728796
Gene Chromosome Location
Chr.9: 136796338-136807741 on Build GRCh38
Transcripts
NM_001039374.4 NR_024580.1

Gene Symbol
CCDC183-AS1
Entrez Gene ID
100131193
Gene Name
CCDC183 antisense RNA 1
Gene Aliases
KIAA1984-AS1
UniGene
Hs.734812
Gene Chromosome Location
Chr.9: 136803927-136808848 on Build GRCh38
Transcripts
NM_001039374.4 NR_024580.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6473036 c.793-5A>C p.? Chr.9: 136804757-136804757 on Build GRCh38
1497016 c.814C>A p.P272T Chr.9: 136804783-136804783 on Build GRCh38
5518936 c.847+7G>A p.? Chr.9: 136804823-136804823 on Build GRCh38
5908922 c.818C>A p.S273* Chr.9: 136804787-136804787 on Build GRCh38
5573312 c.793-1G>A p.? Chr.9: 136804761-136804761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373434330 Chr.9: 136804810-136804810 on Build GRCh38
rs924810471 Chr.9: 136804918-136804918 on Build GRCh38
rs371973312 Chr.9: 136804788-136804788 on Build GRCh38
rs948790825 Chr.9: 136804969-136804969 on Build GRCh38
rs754204961 Chr.9: 136804839-136804839 on Build GRCh38
rs546742333 Chr.9: 136804889-136804889 on Build GRCh38
rs202046982 Chr.9: 136804782-136804782 on Build GRCh38
rs530143489 Chr.9: 136804822-136804822 on Build GRCh38
rs759300237 Chr.9: 136804793-136804793 on Build GRCh38
rs1022357587 Chr.9: 136804848-136804848 on Build GRCh38
rs892983412 Chr.9: 136804878-136804878 on Build GRCh38
rs750587673 Chr.9: 136804820-136804820 on Build GRCh38
rs770637747 Chr.9: 136804791-136804791 on Build GRCh38
rs992961499 Chr.9: 136804938-136804938 on Build GRCh38
rs375518734 Chr.9: 136804894-136804894 on Build GRCh38
rs1017521422 Chr.9: 136804966-136804966 on Build GRCh38
rs762951463 Chr.9: 136804806-136804806 on Build GRCh38
rs755438072 Chr.9: 136804854-136804854 on Build GRCh38
rs948422011 Chr.9: 136804808-136804808 on Build GRCh38
rs989166398 Chr.9: 136804837-136804837 on Build GRCh38
rs758965362 Chr.9: 136804850-136804850 on Build GRCh38
rs375083486 Chr.9: 136804864-136804864 on Build GRCh38
rs915998928 Chr.9: 136804953-136804953 on Build GRCh38
rs200125241 Chr.9: 136804803-136804803 on Build GRCh38
rs367971685 Chr.9: 136804787-136804787 on Build GRCh38
rs753092206 Chr.9: 136804849-136804849 on Build GRCh38
rs769654558 Chr.9: 136804802-136804802 on Build GRCh38
rs755261114 Chr.9: 136804840-136804840 on Build GRCh38
rs969013086 Chr.9: 136804886-136804886 on Build GRCh38
rs964397764 Chr.9: 136804831-136804831 on Build GRCh38
rs767878379 Chr.9: 136804925-136804925 on Build GRCh38
rs200940788 Chr.9: 136804792-136804792 on Build GRCh38
rs998625989 Chr.9: 136804859-136804859 on Build GRCh38
rs773304728 Chr.9: 136804917-136804918 on Build GRCh38
rs779341699 Chr.9: 136804841-136804841 on Build GRCh38
rs890269507 Chr.9: 136804799-136804799 on Build GRCh38
rs766918598 Chr.9: 136804910-136804910 on Build GRCh38
rs376050386 Chr.9: 136804783-136804783 on Build GRCh38
rs766745692 Chr.9: 136804823-136804823 on Build GRCh38

View Full Product Details