Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 186251756-186252013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLKB1
Entrez Gene ID
3818
Gene Name
kallikrein B1
Gene Aliases
KLK3, PKK, PKKD, PPK
UniGene
Hs.237642
Gene Chromosome Location
Chr.4: 186215714-186258477 on Build GRCh38
Transcripts
NM_000892.4 NM_001318394.1 NM_001318396.1 XM_011531930.2 XM_017008181.1 XM_017008182.1 XM_017008183.1 XM_017008184.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3602575 c.1055C>T p.S352F Chr.4: 186251772-186251772 on Build GRCh38
4963238 c.1083G>A p.A361A Chr.4: 186251800-186251800 on Build GRCh38
3602577 c.1116G>A p.L372L Chr.4: 186251833-186251833 on Build GRCh38
4123822 c.1082C>T p.A361V Chr.4: 186251799-186251799 on Build GRCh38
6166873 c.1044C>G p.F348L Chr.4: 186251761-186251761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs536059007 Chr.4: 186251817-186251817 on Build GRCh38
rs267600112 Chr.4: 186251794-186251794 on Build GRCh38
rs777290954 Chr.4: 186251884-186251884 on Build GRCh38
rs4253301 Chr.4: 186251858-186251858 on Build GRCh38
rs930534202 Chr.4: 186251968-186251968 on Build GRCh38
rs764480907 Chr.4: 186251831-186251831 on Build GRCh38
rs770781234 Chr.4: 186251895-186251895 on Build GRCh38
rs142587342 Chr.4: 186251964-186251964 on Build GRCh38
rs778767902 Chr.4: 186251976-186251976 on Build GRCh38
rs567160749 Chr.4: 186251800-186251800 on Build GRCh38
rs376959569 Chr.4: 186251980-186251980 on Build GRCh38
rs553106583 Chr.4: 186251827-186251827 on Build GRCh38
rs779906698 Chr.4: 186251914-186251914 on Build GRCh38
rs1044527329 Chr.4: 186251802-186251802 on Build GRCh38
rs760888127 Chr.4: 186251820-186251820 on Build GRCh38
rs774212017 Chr.4: 186251985-186251986 on Build GRCh38
rs765805024 Chr.4: 186251840-186251840 on Build GRCh38
rs780873341 Chr.4: 186251870-186251870 on Build GRCh38
rs750899027 Chr.4: 186251850-186251850 on Build GRCh38
rs112774511 Chr.4: 186251882-186251882 on Build GRCh38
rs747716379 Chr.4: 186251799-186251799 on Build GRCh38
rs375447593 Chr.4: 186251855-186251855 on Build GRCh38
rs4253380 Chr.4: 186251886-186251886 on Build GRCh38
rs112693910 Chr.4: 186251865-186251865 on Build GRCh38
rs762030727 Chr.4: 186251836-186251836 on Build GRCh38
rs148399602 Chr.4: 186251876-186251876 on Build GRCh38
rs938703817 Chr.4: 186251828-186251828 on Build GRCh38
rs1028071307 Chr.4: 186251990-186251990 on Build GRCh38
rs749215436 Chr.4: 186251814-186251814 on Build GRCh38
rs996619217 Chr.4: 186251974-186251974 on Build GRCh38
rs770894352 Chr.4: 186251979-186251980 on Build GRCh38
rs368165642 Chr.4: 186251857-186251857 on Build GRCh38
rs1049375110 Chr.4: 186251978-186251978 on Build GRCh38
rs139190582 Chr.4: 186251815-186251815 on Build GRCh38
rs4253379 Chr.4: 186251789-186251789 on Build GRCh38
rs572857859 Chr.4: 186251833-186251833 on Build GRCh38

View Full Product Details