Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44399788-44400013 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPATCH8
Entrez Gene ID
23131
Gene Name
G-patch domain containing 8
Gene Aliases
GPATC8, KIAA0553
UniGene
Hs.463129
Gene Chromosome Location
Chr.17: 44395277-44503602 on Build GRCh38
Transcripts
NM_001002909.3 NM_001304939.1 NM_001304940.1 NM_001304941.1 NM_001304942.1 NM_001304943.1 XM_011524557.1 XM_011524558.1 XM_011524559.2 XM_011524561.2 XM_017024372.1 XM_017024373.1 XM_017024374.1 XM_017024375.1 XM_017024376.1 XM_017024377.1 XM_017024378.1 XM_017024379.1 XM_017024380.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1383675 c.2146C>T p.R716* Chr.17: 44399931-44399931 on Build GRCh38
4995750 c.2284G>A p.E762K Chr.17: 44399793-44399793 on Build GRCh38
4611745 c.2207delC p.P736fs*373 Chr.17: 44399870-44399870 on Build GRCh38
6523586 c.2116G>T p.E706* Chr.17: 44399961-44399961 on Build GRCh38
5747863 c.2105C>T p.A702V Chr.17: 44399972-44399972 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778323094 Chr.17: 44399853-44399853 on Build GRCh38
rs200958648 Chr.17: 44399972-44399972 on Build GRCh38
rs775941625 Chr.17: 44399847-44399847 on Build GRCh38
rs770210247 Chr.17: 44399835-44399835 on Build GRCh38
rs749352671 Chr.17: 44399945-44399945 on Build GRCh38
rs764970937 Chr.17: 44399920-44399920 on Build GRCh38
rs779037030 Chr.17: 44399958-44399958 on Build GRCh38
rs763495479 Chr.17: 44399983-44399983 on Build GRCh38
rs138548294 Chr.17: 44399991-44399991 on Build GRCh38
rs1033990075 Chr.17: 44399990-44399990 on Build GRCh38
rs747355946 Chr.17: 44399854-44399854 on Build GRCh38
rs982373110 Chr.17: 44399989-44399989 on Build GRCh38
rs751562055 Chr.17: 44399919-44399919 on Build GRCh38
rs369496644 Chr.17: 44399930-44399930 on Build GRCh38
rs369762319 Chr.17: 44399992-44399992 on Build GRCh38
rs752039281 Chr.17: 44399924-44399924 on Build GRCh38
rs762966534 Chr.17: 44399866-44399866 on Build GRCh38
rs902117611 Chr.17: 44399915-44399915 on Build GRCh38
rs750013557 Chr.17: 44399833-44399833 on Build GRCh38
rs374161973 Chr.17: 44399964-44399964 on Build GRCh38
rs775813278 Chr.17: 44399875-44399875 on Build GRCh38
rs750852938 Chr.17: 44399936-44399936 on Build GRCh38
rs780046555 Chr.17: 44399940-44399940 on Build GRCh38
rs757899271 Chr.17: 44399861-44399861 on Build GRCh38
rs769712512 Chr.17: 44399871-44399871 on Build GRCh38
rs778513845 Chr.17: 44399837-44399839 on Build GRCh38
rs543950617 Chr.17: 44399937-44399937 on Build GRCh38
rs1002066011 Chr.17: 44399979-44399979 on Build GRCh38
rs754836748 Chr.17: 44399956-44399956 on Build GRCh38
rs772968706 Chr.17: 44399900-44399900 on Build GRCh38
rs749550846 Chr.17: 44399878-44399878 on Build GRCh38
rs773236992 Chr.17: 44399965-44399965 on Build GRCh38
rs763836264 Chr.17: 44399811-44399811 on Build GRCh38
rs762758258 Chr.17: 44399810-44399810 on Build GRCh38
rs760257617 Chr.17: 44399903-44399903 on Build GRCh38
rs1020367735 Chr.17: 44399923-44399923 on Build GRCh38
rs201098137 Chr.17: 44399891-44399891 on Build GRCh38
rs565338842 Chr.17: 44399962-44399962 on Build GRCh38
rs758869547 Chr.17: 44399852-44399852 on Build GRCh38
rs781733057 Chr.17: 44399934-44399934 on Build GRCh38
rs181814284 Chr.17: 44399907-44399907 on Build GRCh38
rs767340798 Chr.17: 44399828-44399828 on Build GRCh38
rs140576995 Chr.17: 44399834-44399834 on Build GRCh38
rs768840434 Chr.17: 44399879-44399879 on Build GRCh38
rs760573373 Chr.17: 44399969-44399969 on Build GRCh38
rs774185994 Chr.17: 44399881-44399881 on Build GRCh38
rs774100263 Chr.17: 44399813-44399813 on Build GRCh38
rs753386957 Chr.17: 44399848-44399848 on Build GRCh38
rs1043832034 Chr.17: 44399836-44399836 on Build GRCh38
rs74823475 Chr.17: 44399914-44399914 on Build GRCh38
rs3744426 Chr.17: 44399821-44399821 on Build GRCh38
rs868300404 Chr.17: 44399946-44399946 on Build GRCh38
rs1008320991 Chr.17: 44399929-44399929 on Build GRCh38
rs776338526 Chr.17: 44399981-44399981 on Build GRCh38
rs745540283 Chr.17: 44399964-44399965 on Build GRCh38
rs781547870 Chr.17: 44399864-44399864 on Build GRCh38

View Full Product Details