Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78891820-78892071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP295NL
Entrez Gene ID
100653515
Gene Name
CEP295 N-terminal like
Gene Aliases
DDC8, KIAA1731NL
UniGene
Hs.734636
Gene Chromosome Location
Chr.17: 78890580-78903217 on Build GRCh38
Transcripts
NM_001243540.1 NM_001243541.1 NM_003255.4

Gene Symbol
TIMP2
Entrez Gene ID
7077
Gene Name
TIMP metallopeptidase inhibitor 2
Gene Aliases
CSC-21K, DDC8
UniGene
Hs.633514
Gene Chromosome Location
Chr.17: 78852977-78925390 on Build GRCh38
Transcripts
NM_001243540.1 NM_001243541.1 NM_003255.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4936784 c.553G>T p.G185C Chr.17: 78891951-78891951 on Build GRCh38
4433049 c.635G>C p.R212P Chr.17: 78891869-78891869 on Build GRCh38
3378374 c.479C>G p.S160* Chr.17: 78892025-78892025 on Build GRCh38
6046402 c.678G>A p.R226R Chr.17: 78891826-78891826 on Build GRCh38
4958935 c.555C>T p.G185G Chr.17: 78891949-78891949 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769548889 Chr.17: 78891847-78891847 on Build GRCh38
rs756676057 Chr.17: 78891875-78891875 on Build GRCh38
rs949931835 Chr.17: 78891885-78891885 on Build GRCh38
rs773913971 Chr.17: 78891905-78891905 on Build GRCh38
rs754343868 Chr.17: 78891902-78891902 on Build GRCh38
rs118048577 Chr.17: 78892009-78892009 on Build GRCh38
rs767256502 Chr.17: 78891921-78891921 on Build GRCh38
rs758910481 Chr.17: 78891914-78891914 on Build GRCh38
rs935785248 Chr.17: 78891870-78891870 on Build GRCh38
rs777572406 Chr.17: 78892048-78892048 on Build GRCh38
rs8077028 Chr.17: 78891959-78891959 on Build GRCh38
rs781520503 Chr.17: 78892014-78892014 on Build GRCh38
rs1008258771 Chr.17: 78891955-78891955 on Build GRCh38
rs769950393 Chr.17: 78892032-78892032 on Build GRCh38
rs755011713 Chr.17: 78892012-78892012 on Build GRCh38
rs368596724 Chr.17: 78892023-78892023 on Build GRCh38
rs770811442 Chr.17: 78891880-78891880 on Build GRCh38
rs748045323 Chr.17: 78891843-78891843 on Build GRCh38
rs1029098852 Chr.17: 78891997-78891997 on Build GRCh38
rs1005254777 Chr.17: 78891957-78891957 on Build GRCh38
rs140599940 Chr.17: 78891995-78891995 on Build GRCh38
rs546192846 Chr.17: 78891942-78891942 on Build GRCh38
rs925617526 Chr.17: 78891865-78891865 on Build GRCh38
rs752287126 Chr.17: 78891935-78891935 on Build GRCh38
rs180840662 Chr.17: 78891879-78891879 on Build GRCh38
rs1037234759 Chr.17: 78891874-78891874 on Build GRCh38
rs773186542 Chr.17: 78891848-78891848 on Build GRCh38
rs535553743 Chr.17: 78892044-78892044 on Build GRCh38
rs879179049 Chr.17: 78891920-78891920 on Build GRCh38
rs865931331 Chr.17: 78891908-78891908 on Build GRCh38
rs1049127123 Chr.17: 78891936-78891936 on Build GRCh38
rs748352312 Chr.17: 78892031-78892031 on Build GRCh38
rs571823006 Chr.17: 78891962-78891962 on Build GRCh38
rs905804280 Chr.17: 78891931-78891931 on Build GRCh38
rs867893339 Chr.17: 78891909-78891909 on Build GRCh38
rs889445682 Chr.17: 78891950-78891950 on Build GRCh38
rs959354980 Chr.17: 78892016-78892016 on Build GRCh38
rs1011091870 Chr.17: 78891976-78891976 on Build GRCh38
rs941212699 Chr.17: 78891873-78891873 on Build GRCh38
rs564686703 Chr.17: 78892003-78892003 on Build GRCh38
rs1021106466 Chr.17: 78891994-78891994 on Build GRCh38
rs925503035 Chr.17: 78892049-78892049 on Build GRCh38

View Full Product Details