Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 36602091-36602350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MRM1
Entrez Gene ID
79922
Gene Name
mitochondrial rRNA methyltransferase 1
Gene Aliases
-
UniGene
Hs.194864
Gene Chromosome Location
Chr.17: 36601581-36634698 on Build GRCh38
Transcripts
NM_024864.4 XM_005257694.4 XM_011525275.2 XM_011525276.2 XM_017025112.1

SNP Detail

SNP ID Chromosome Location
rs548319487 Chr.17: 36602214-36602214 on Build GRCh38
rs775359295 Chr.17: 36602306-36602306 on Build GRCh38
rs200505410 Chr.17: 36602286-36602286 on Build GRCh38
rs545927735 Chr.17: 36602324-36602324 on Build GRCh38
rs753771681 Chr.17: 36602120-36602120 on Build GRCh38
rs780671818 Chr.17: 36602256-36602256 on Build GRCh38
rs755716309 Chr.17: 36602255-36602255 on Build GRCh38
rs777568515 Chr.17: 36602236-36602236 on Build GRCh38
rs756863939 Chr.17: 36602249-36602249 on Build GRCh38
rs199548537 Chr.17: 36602144-36602144 on Build GRCh38
rs143242016 Chr.17: 36602279-36602279 on Build GRCh38
rs780693186 Chr.17: 36602251-36602251 on Build GRCh38
rs114566242 Chr.17: 36602211-36602211 on Build GRCh38
rs769179096 Chr.17: 36602182-36602182 on Build GRCh38
rs765158355 Chr.17: 36602216-36602216 on Build GRCh38
rs761960719 Chr.17: 36602191-36602191 on Build GRCh38
rs372983796 Chr.17: 36602310-36602310 on Build GRCh38
rs1045586368 Chr.17: 36602262-36602262 on Build GRCh38
rs745357242 Chr.17: 36602254-36602254 on Build GRCh38
rs935782728 Chr.17: 36602205-36602205 on Build GRCh38
rs758168462 Chr.17: 36602233-36602233 on Build GRCh38
rs773554294 Chr.17: 36602291-36602291 on Build GRCh38
rs948430654 Chr.17: 36602234-36602234 on Build GRCh38
rs752672429 Chr.17: 36602220-36602220 on Build GRCh38
rs918540276 Chr.17: 36602152-36602152 on Build GRCh38
rs771708633 Chr.17: 36602330-36602331 on Build GRCh38
rs569167285 Chr.17: 36602137-36602137 on Build GRCh38
rs376237753 Chr.17: 36602322-36602322 on Build GRCh38
rs757133378 Chr.17: 36602323-36602323 on Build GRCh38
rs201462793 Chr.17: 36602153-36602153 on Build GRCh38
rs763402009 Chr.17: 36602170-36602170 on Build GRCh38
rs112713872 Chr.17: 36602186-36602186 on Build GRCh38
rs758256753 Chr.17: 36602138-36602138 on Build GRCh38
rs777374487 Chr.17: 36602143-36602143 on Build GRCh38
rs778716807 Chr.17: 36602126-36602126 on Build GRCh38
rs376476399 Chr.17: 36602203-36602203 on Build GRCh38
rs1000205771 Chr.17: 36602331-36602332 on Build GRCh38
rs568437431 Chr.17: 36602261-36602261 on Build GRCh38
rs767540749 Chr.17: 36602112-36602112 on Build GRCh38
rs766494977 Chr.17: 36602303-36602303 on Build GRCh38
rs774863553 Chr.17: 36602267-36602267 on Build GRCh38
rs78943308 Chr.17: 36602169-36602169 on Build GRCh38
rs774689858 Chr.17: 36602190-36602190 on Build GRCh38
rs780596708 Chr.17: 36602145-36602145 on Build GRCh38
rs749857579 Chr.17: 36602259-36602259 on Build GRCh38
rs138258458 Chr.17: 36602314-36602314 on Build GRCh38
rs751185835 Chr.17: 36602245-36602245 on Build GRCh38
rs748347544 Chr.17: 36602271-36602271 on Build GRCh38

View Full Product Details