Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13100976-13101205 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYL1
Entrez Gene ID
4066
Gene Name
LYL1, basic helix-loop-helix family member
Gene Aliases
bHLHa18
UniGene
Hs.46446
Gene Chromosome Location
Chr.19: 13099028-13103160 on Build GRCh38
Transcripts
NM_005583.4 XM_005259912.3 XM_017026816.1 XM_017026817.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3756356 c.165G>A p.R55R Chr.19: 13101007-13101007 on Build GRCh38
5493846 c.162delC p.R55fs*8 Chr.19: 13101010-13101010 on Build GRCh38
991576 c.129G>T p.E43D Chr.19: 13101043-13101043 on Build GRCh38
5416997 c.186G>C p.V62V Chr.19: 13100986-13100986 on Build GRCh38
6397433 c.124G>A p.V42M Chr.19: 13101048-13101048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748749696 Chr.19: 13101015-13101015 on Build GRCh38
rs868596235 Chr.19: 13101158-13101158 on Build GRCh38
rs764330099 Chr.19: 13101109-13101109 on Build GRCh38
rs746763875 Chr.19: 13101097-13101097 on Build GRCh38
rs763217304 Chr.19: 13101052-13101052 on Build GRCh38
rs767978111 Chr.19: 13101127-13101127 on Build GRCh38
rs778541539 Chr.19: 13101081-13101081 on Build GRCh38
rs756741072 Chr.19: 13101078-13101078 on Build GRCh38
rs374243329 Chr.19: 13101073-13101073 on Build GRCh38
rs770721427 Chr.19: 13101101-13101101 on Build GRCh38
rs762162133 Chr.19: 13101124-13101124 on Build GRCh38
rs991135036 Chr.19: 13101178-13101178 on Build GRCh38
rs1036009996 Chr.19: 13101164-13101164 on Build GRCh38
rs1032028833 Chr.19: 13101136-13101136 on Build GRCh38
rs777309732 Chr.19: 13101092-13101092 on Build GRCh38
rs756964717 Chr.19: 13101182-13101182 on Build GRCh38
rs781772639 Chr.19: 13101008-13101008 on Build GRCh38
rs765299335 Chr.19: 13101040-13101040 on Build GRCh38
rs745795686 Chr.19: 13101104-13101104 on Build GRCh38
rs891384752 Chr.19: 13101024-13101024 on Build GRCh38
rs764263445 Chr.19: 13101053-13101053 on Build GRCh38
rs867932185 Chr.19: 13101159-13101159 on Build GRCh38
rs371499313 Chr.19: 13101163-13101163 on Build GRCh38
rs201786409 Chr.19: 13101064-13101064 on Build GRCh38
rs750865544 Chr.19: 13101131-13101131 on Build GRCh38
rs752197880 Chr.19: 13101153-13101153 on Build GRCh38
rs775208969 Chr.19: 13101174-13101174 on Build GRCh38
rs554767683 Chr.19: 13101144-13101144 on Build GRCh38
rs774440875 Chr.19: 13101110-13101110 on Build GRCh38
rs756655151 Chr.19: 13101139-13101139 on Build GRCh38
rs576500504 Chr.19: 13101183-13101183 on Build GRCh38
rs781736635 Chr.19: 13101072-13101072 on Build GRCh38
rs117072928 Chr.19: 13101029-13101029 on Build GRCh38
rs971615157 Chr.19: 13101122-13101122 on Build GRCh38
rs768286348 Chr.19: 13101016-13101016 on Build GRCh38
rs775611060 Chr.19: 13101105-13101105 on Build GRCh38
rs778631540 Chr.19: 13101017-13101017 on Build GRCh38
rs762975049 Chr.19: 13101106-13101106 on Build GRCh38
rs375885774 Chr.19: 13101181-13101181 on Build GRCh38
rs751920976 Chr.19: 13100999-13100999 on Build GRCh38
rs770857160 Chr.19: 13101035-13101035 on Build GRCh38
rs777421599 Chr.19: 13101175-13101175 on Build GRCh38
rs760414229 Chr.19: 13101033-13101033 on Build GRCh38
rs771711636 Chr.19: 13101091-13101091 on Build GRCh38
rs1056435720 Chr.19: 13101082-13101082 on Build GRCh38
rs1047732287 Chr.19: 13101065-13101065 on Build GRCh38
rs759547940 Chr.19: 13101038-13101038 on Build GRCh38
rs141274358 Chr.19: 13101023-13101023 on Build GRCh38
rs747649669 Chr.19: 13101088-13101088 on Build GRCh38
rs901295130 Chr.19: 13101051-13101051 on Build GRCh38
rs151073349 Chr.19: 13101007-13101007 on Build GRCh38

View Full Product Details