Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 166383-166644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HBM
Entrez Gene ID
3042
Gene Name
hemoglobin subunit mu
Gene Aliases
HBAP2, HBK
UniGene
Hs.647389
Gene Chromosome Location
Chr.16: 165974-166768 on Build GRCh38
Transcripts
NM_001003938.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
968483 c.301C>A p.L101I Chr.16: 166583-166583 on Build GRCh38
7281669 c.233C>T p.A78V Chr.16: 166408-166408 on Build GRCh38
4128809 c.312T>C p.C104C Chr.16: 166594-166594 on Build GRCh38
4766485 c.296C>T p.P99L Chr.16: 166471-166471 on Build GRCh38
3749354 c.246G>T p.P82P Chr.16: 166421-166421 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750003435 Chr.16: 166624-166624 on Build GRCh38
rs777251866 Chr.16: 166440-166440 on Build GRCh38
rs2858936 Chr.16: 166517-166517 on Build GRCh38
rs756737546 Chr.16: 166537-166537 on Build GRCh38
rs755549539 Chr.16: 166518-166518 on Build GRCh38
rs118054833 Chr.16: 166421-166421 on Build GRCh38
rs933023405 Chr.16: 166413-166413 on Build GRCh38
rs369709218 Chr.16: 166516-166516 on Build GRCh38
rs765565200 Chr.16: 166519-166519 on Build GRCh38
rs745625020 Chr.16: 166479-166479 on Build GRCh38
rs763685109 Chr.16: 166597-166597 on Build GRCh38
rs766862068 Chr.16: 166506-166506 on Build GRCh38
rs750593544 Chr.16: 166409-166409 on Build GRCh38
rs769815020 Chr.16: 166544-166544 on Build GRCh38
rs745757790 Chr.16: 166543-166543 on Build GRCh38
rs758095270 Chr.16: 166438-166438 on Build GRCh38
rs982314539 Chr.16: 166403-166403 on Build GRCh38
rs756971365 Chr.16: 166602-166602 on Build GRCh38
rs760769781 Chr.16: 166563-166563 on Build GRCh38
rs1016763664 Chr.16: 166554-166554 on Build GRCh38
rs758040184 Chr.16: 166523-166523 on Build GRCh38
rs755322486 Chr.16: 166427-166427 on Build GRCh38
rs756687997 Chr.16: 166420-166420 on Build GRCh38
rs772366255 Chr.16: 166555-166555 on Build GRCh38
rs773146864 Chr.16: 166571-166573 on Build GRCh38
rs762100374 Chr.16: 166489-166489 on Build GRCh38
rs776444946 Chr.16: 166457-166457 on Build GRCh38
rs773592456 Chr.16: 166562-166562 on Build GRCh38
rs770832988 Chr.16: 166448-166448 on Build GRCh38
rs765800320 Chr.16: 166583-166583 on Build GRCh38
rs746588318 Chr.16: 166442-166442 on Build GRCh38
rs548436387 Chr.16: 166488-166488 on Build GRCh38
rs967258464 Chr.16: 166564-166564 on Build GRCh38
rs777405838 Chr.16: 166524-166524 on Build GRCh38
rs530193442 Chr.16: 166434-166434 on Build GRCh38
rs752991428 Chr.16: 166593-166593 on Build GRCh38
rs558533181 Chr.16: 166532-166532 on Build GRCh38
rs1038600092 Chr.16: 166459-166459 on Build GRCh38
rs759947842 Chr.16: 166579-166579 on Build GRCh38
rs754176161 Chr.16: 166424-166424 on Build GRCh38
rs898720724 Chr.16: 166463-166463 on Build GRCh38
rs867591404 Chr.16: 166443-166443 on Build GRCh38
rs775425967 Chr.16: 166546-166546 on Build GRCh38
rs767922500 Chr.16: 166493-166493 on Build GRCh38
rs776821733 Chr.16: 166577-166577 on Build GRCh38
rs781140892 Chr.16: 166542-166542 on Build GRCh38
rs780541464 Chr.16: 166610-166610 on Build GRCh38
rs767687114 Chr.16: 166400-166400 on Build GRCh38
rs773363094 Chr.16: 166497-166497 on Build GRCh38
rs760686044 Chr.16: 166503-166503 on Build GRCh38
rs547802002 Chr.16: 166412-166412 on Build GRCh38
rs770885537 Chr.16: 166567-166567 on Build GRCh38
rs537357463 Chr.16: 166528-166528 on Build GRCh38
rs745655084 Chr.16: 166582-166583 on Build GRCh38
rs373124053 Chr.16: 166482-166482 on Build GRCh38
rs141494605 Chr.16: 166594-166594 on Build GRCh38
rs893523671 Chr.16: 166436-166436 on Build GRCh38
rs770468517 Chr.16: 166518-166519 on Build GRCh38
rs143725294 Chr.16: 166601-166601 on Build GRCh38

View Full Product Details