Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35939434-35939683 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRFN3
Entrez Gene ID
79414
Gene Name
leucine rich repeat and fibronectin type III domain containing 3
Gene Aliases
FIGLER1, SALM4
UniGene
Hs.143792
Gene Chromosome Location
Chr.19: 35937120-35945195 on Build GRCh38
Transcripts
NM_024509.1 XM_017027302.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6702881 c.119C>G p.P40R Chr.19: 35939544-35939544 on Build GRCh38
3532848 c.141G>A p.G47G Chr.19: 35939566-35939566 on Build GRCh38
3532847 c.62A>T p.Q21L Chr.19: 35939487-35939487 on Build GRCh38
3532846 c.50C>T p.S17L Chr.19: 35939475-35939475 on Build GRCh38
6702879 c.88C>T p.R30C Chr.19: 35939513-35939513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756529001 Chr.19: 35939509-35939509 on Build GRCh38
rs145812907 Chr.19: 35939572-35939572 on Build GRCh38
rs539638458 Chr.19: 35939523-35939523 on Build GRCh38
rs748369999 Chr.19: 35939454-35939454 on Build GRCh38
rs755492282 Chr.19: 35939514-35939514 on Build GRCh38
rs754225734 Chr.19: 35939581-35939581 on Build GRCh38
rs765803134 Chr.19: 35939651-35939651 on Build GRCh38
rs377765613 Chr.19: 35939611-35939611 on Build GRCh38
rs139243712 Chr.19: 35939456-35939456 on Build GRCh38
rs763750148 Chr.19: 35939503-35939503 on Build GRCh38
rs956854561 Chr.19: 35939493-35939493 on Build GRCh38
rs770728711 Chr.19: 35939538-35939538 on Build GRCh38
rs573957542 Chr.19: 35939627-35939627 on Build GRCh38
rs755450034 Chr.19: 35939582-35939582 on Build GRCh38
rs373149661 Chr.19: 35939602-35939602 on Build GRCh38
rs773788876 Chr.19: 35939639-35939639 on Build GRCh38
rs769725843 Chr.19: 35939551-35939551 on Build GRCh38
rs766689702 Chr.19: 35939510-35939510 on Build GRCh38
rs146159415 Chr.19: 35939478-35939478 on Build GRCh38
rs745806923 Chr.19: 35939546-35939546 on Build GRCh38
rs371234713 Chr.19: 35939623-35939623 on Build GRCh38
rs760535700 Chr.19: 35939461-35939461 on Build GRCh38
rs763603932 Chr.19: 35939654-35939654 on Build GRCh38
rs912468422 Chr.19: 35939603-35939603 on Build GRCh38
rs376224731 Chr.19: 35939554-35939554 on Build GRCh38
rs762524415 Chr.19: 35939482-35939482 on Build GRCh38
rs776341900 Chr.19: 35939539-35939539 on Build GRCh38
rs776661833 Chr.19: 35939473-35939473 on Build GRCh38
rs760004767 Chr.19: 35939649-35939649 on Build GRCh38
rs373172486 Chr.19: 35939657-35939657 on Build GRCh38
rs140156601 Chr.19: 35939524-35939524 on Build GRCh38
rs947789538 Chr.19: 35939522-35939522 on Build GRCh38
rs750737092 Chr.19: 35939506-35939506 on Build GRCh38
rs34933126 Chr.19: 35939466-35939466 on Build GRCh38
rs766749662 Chr.19: 35939575-35939575 on Build GRCh38
rs752779379 Chr.19: 35939600-35939600 on Build GRCh38
rs764222042 Chr.19: 35939655-35939655 on Build GRCh38
rs773078781 Chr.19: 35939460-35939460 on Build GRCh38
rs879310875 Chr.19: 35939593-35939593 on Build GRCh38
rs368952403 Chr.19: 35939601-35939601 on Build GRCh38
rs373273430 Chr.19: 35939475-35939475 on Build GRCh38
rs768128608 Chr.19: 35939638-35939638 on Build GRCh38
rs757480500 Chr.19: 35939604-35939604 on Build GRCh38
rs142556345 Chr.19: 35939644-35939644 on Build GRCh38
rs754345820 Chr.19: 35939513-35939513 on Build GRCh38
rs200309945 Chr.19: 35939626-35939626 on Build GRCh38
rs1037268851 Chr.19: 35939612-35939612 on Build GRCh38
rs2242590 Chr.19: 35939563-35939563 on Build GRCh38
rs751658979 Chr.19: 35939656-35939656 on Build GRCh38
rs762471385 Chr.19: 35939555-35939555 on Build GRCh38
rs764783178 Chr.19: 35939477-35939477 on Build GRCh38
rs746742249 Chr.19: 35939533-35939533 on Build GRCh38
rs753224798 Chr.19: 35939652-35939652 on Build GRCh38
rs779133099 Chr.19: 35939517-35939517 on Build GRCh38
rs776999907 Chr.19: 35939648-35939648 on Build GRCh38
rs745661591 Chr.19: 35939619-35939619 on Build GRCh38
rs774019455 Chr.19: 35939566-35939566 on Build GRCh38
rs1027061018 Chr.19: 35939504-35939504 on Build GRCh38
rs148437277 Chr.19: 35939592-35939592 on Build GRCh38

View Full Product Details