Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 157008857-157009114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MNX1
Entrez Gene ID
3110
Gene Name
motor neuron and pancreas homeobox 1
Gene Aliases
HB9, HLXB9, HOXHB9, SCRA1
UniGene
Hs.37035
Gene Chromosome Location
Chr.7: 157004853-157010653 on Build GRCh38
Transcripts
NM_001165255.1 NM_005515.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7252696 c.47C>A p.A16D Chr.7: 157009006-157009006 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs977507967 Chr.7: 157008977-157008977 on Build GRCh38
rs919788941 Chr.7: 157009023-157009023 on Build GRCh38
rs544000821 Chr.7: 157008975-157008975 on Build GRCh38
rs1046887170 Chr.7: 157009073-157009073 on Build GRCh38
rs373686150 Chr.7: 157009005-157009005 on Build GRCh38
rs546454726 Chr.7: 157009087-157009087 on Build GRCh38
rs1031632603 Chr.7: 157008905-157008905 on Build GRCh38
rs924694736 Chr.7: 157008984-157008984 on Build GRCh38
rs923143766 Chr.7: 157008939-157008939 on Build GRCh38
rs77813062 Chr.7: 157008967-157008967 on Build GRCh38
rs781204492 Chr.7: 157008950-157008950 on Build GRCh38
rs959183784 Chr.7: 157008951-157008951 on Build GRCh38
rs1002551728 Chr.7: 157009091-157009091 on Build GRCh38
rs151220659 Chr.7: 157008993-157008993 on Build GRCh38
rs779610709 Chr.7: 157008908-157008908 on Build GRCh38
rs576582946 Chr.7: 157008963-157008963 on Build GRCh38
rs775180588 Chr.7: 157008969-157008969 on Build GRCh38
rs1013102648 Chr.7: 157008957-157008957 on Build GRCh38
rs777387160 Chr.7: 157008892-157008892 on Build GRCh38
rs968758575 Chr.7: 157008880-157008880 on Build GRCh38
rs768385251 Chr.7: 157008986-157008986 on Build GRCh38
rs752801081 Chr.7: 157009094-157009094 on Build GRCh38
rs898267634 Chr.7: 157008893-157008893 on Build GRCh38
rs1025386474 Chr.7: 157008902-157008902 on Build GRCh38
rs995280729 Chr.7: 157008900-157008900 on Build GRCh38
rs1039916710 Chr.7: 157008881-157008881 on Build GRCh38
rs757084352 Chr.7: 157008980-157008980 on Build GRCh38
rs931127399 Chr.7: 157009034-157009034 on Build GRCh38
rs78596384 Chr.7: 157009006-157009006 on Build GRCh38
rs530397959 Chr.7: 157008921-157008921 on Build GRCh38
rs559932872 Chr.7: 157008877-157008877 on Build GRCh38
rs146062190 Chr.7: 157008992-157008992 on Build GRCh38
rs886809347 Chr.7: 157008906-157008906 on Build GRCh38
rs1013195122 Chr.7: 157009044-157009045 on Build GRCh38
rs753593706 Chr.7: 157009045-157009046 on Build GRCh38
rs977497988 Chr.7: 157008886-157008886 on Build GRCh38
rs756417339 Chr.7: 157009022-157009025 on Build GRCh38
rs765274730 Chr.7: 157009064-157009064 on Build GRCh38

View Full Product Details