Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 96355138-96355411 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OPALIN
Entrez Gene ID
93377
Gene Name
oligodendrocytic myelin paranodal and inner loop protein
Gene Aliases
HTMP10, TMEM10, TMP10
UniGene
Hs.12449
Gene Chromosome Location
Chr.10: 96343216-96359365 on Build GRCh38
Transcripts
NM_001040102.2 NM_001040103.2 NM_001284320.1 NM_001284321.1 NM_001284322.1 NM_001284323.1 NM_001284324.1 NM_001284326.1 NM_001284327.1 NM_033207.4 NR_104296.1 NR_104297.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4709491 c.18C>T p.N6N Chr.10: 96355276-96355276 on Build GRCh38
4709490 c.29C>T p.P10L Chr.10: 96355265-96355265 on Build GRCh38
6489889 c.29C>A p.P10Q Chr.10: 96355265-96355265 on Build GRCh38
5371417 c.33G>A p.A11A Chr.10: 96355261-96355261 on Build GRCh38
364277 c.25C>G p.L9V Chr.10: 96355269-96355269 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs777752809 Chr.10: 96355310-96355310 on Build GRCh38
rs116374728 Chr.10: 96355380-96355380 on Build GRCh38
rs201844051 Chr.10: 96355273-96355273 on Build GRCh38
rs1036309415 Chr.10: 96355173-96355173 on Build GRCh38
rs1046342163 Chr.10: 96355265-96355265 on Build GRCh38
rs760203194 Chr.10: 96355261-96355261 on Build GRCh38
rs532384592 Chr.10: 96355320-96355320 on Build GRCh38
rs752401220 Chr.10: 96355256-96355256 on Build GRCh38
rs760433820 Chr.10: 96355325-96355325 on Build GRCh38
rs199717457 Chr.10: 96355223-96355223 on Build GRCh38
rs552134701 Chr.10: 96355323-96355323 on Build GRCh38
rs756141366 Chr.10: 96355305-96355305 on Build GRCh38
rs185762163 Chr.10: 96355290-96355290 on Build GRCh38
rs565674602 Chr.10: 96355328-96355328 on Build GRCh38
rs759209578 Chr.10: 96355236-96355236 on Build GRCh38
rs144715395 Chr.10: 96355191-96355191 on Build GRCh38
rs1031338828 Chr.10: 96355332-96355332 on Build GRCh38
rs756577858 Chr.10: 96355357-96355357 on Build GRCh38
rs755193169 Chr.10: 96355283-96355283 on Build GRCh38
rs377257004 Chr.10: 96355176-96355176 on Build GRCh38
rs748386926 Chr.10: 96355301-96355301 on Build GRCh38
rs11188733 Chr.10: 96355299-96355299 on Build GRCh38
rs767345645 Chr.10: 96355247-96355247 on Build GRCh38
rs894604114 Chr.10: 96355249-96355249 on Build GRCh38
rs892809312 Chr.10: 96355376-96355376 on Build GRCh38
rs750903544 Chr.10: 96355263-96355263 on Build GRCh38
rs111883171 Chr.10: 96355264-96355264 on Build GRCh38
rs1184973 Chr.10: 96355317-96355317 on Build GRCh38
rs112566019 Chr.10: 96355381-96355381 on Build GRCh38
rs780978001 Chr.10: 96355326-96355327 on Build GRCh38
rs774522847 Chr.10: 96355237-96355237 on Build GRCh38
rs772004735 Chr.10: 96355324-96355324 on Build GRCh38
rs41306852 Chr.10: 96355262-96355262 on Build GRCh38
rs1026671365 Chr.10: 96355177-96355177 on Build GRCh38
rs527360891 Chr.10: 96355208-96355208 on Build GRCh38
rs751927342 Chr.10: 96355278-96355278 on Build GRCh38
rs774058997 Chr.10: 96355226-96355226 on Build GRCh38
rs930896220 Chr.10: 96355207-96355207 on Build GRCh38
rs773078275 Chr.10: 96355214-96355214 on Build GRCh38
rs764221108 Chr.10: 96355222-96355222 on Build GRCh38

View Full Product Details