Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 143438509-143438740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAB1
Entrez Gene ID
2549
Gene Name
GRB2 associated binding protein 1
Gene Aliases
-
UniGene
Hs.618456
Gene Chromosome Location
Chr.4: 143336830-143474565 on Build GRCh38
Transcripts
NM_002039.3 NM_207123.2 XM_006714167.3 XM_006714168.3 XM_017007966.1 XM_017007967.1 XM_017007968.1 XM_017007969.1 XM_017007970.1 XM_017007971.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4917993 c.1181A>G p.N394S Chr.4: 143438586-143438586 on Build GRCh38
7160858 c.1108G>A p.D370N Chr.4: 143438513-143438513 on Build GRCh38
1051595 c.1150C>T p.R384C Chr.4: 143438555-143438555 on Build GRCh38
6668375 c.1142C>T p.S381L Chr.4: 143438547-143438547 on Build GRCh38
32930 c.1160C>A p.T387N Chr.4: 143438565-143438565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs926695961 Chr.4: 143438673-143438673 on Build GRCh38
rs906537068 Chr.4: 143438627-143438627 on Build GRCh38
rs775653963 Chr.4: 143438633-143438633 on Build GRCh38
rs959651977 Chr.4: 143438655-143438655 on Build GRCh38
rs749679652 Chr.4: 143438554-143438554 on Build GRCh38
rs755764881 Chr.4: 143438548-143438548 on Build GRCh38
rs770216563 Chr.4: 143438704-143438704 on Build GRCh38
rs754537929 Chr.4: 143438679-143438679 on Build GRCh38
rs368426312 Chr.4: 143438608-143438608 on Build GRCh38
rs1037417413 Chr.4: 143438622-143438622 on Build GRCh38
rs888516757 Chr.4: 143438555-143438555 on Build GRCh38
rs911510627 Chr.4: 143438708-143438708 on Build GRCh38
rs55997904 Chr.4: 143438556-143438556 on Build GRCh38
rs375965038 Chr.4: 143438547-143438547 on Build GRCh38
rs1033929073 Chr.4: 143438650-143438650 on Build GRCh38
rs149154042 Chr.4: 143438598-143438598 on Build GRCh38
rs772103709 Chr.4: 143438612-143438612 on Build GRCh38
rs992667873 Chr.4: 143438566-143438566 on Build GRCh38
rs753520226 Chr.4: 143438671-143438671 on Build GRCh38
rs1044267110 Chr.4: 143438652-143438652 on Build GRCh38
rs966423930 Chr.4: 143438690-143438690 on Build GRCh38
rs869217712 Chr.4: 143438714-143438715 on Build GRCh38
rs779332191 Chr.4: 143438558-143438558 on Build GRCh38
rs765983420 Chr.4: 143438541-143438541 on Build GRCh38
rs917184735 Chr.4: 143438649-143438649 on Build GRCh38
rs1011061733 Chr.4: 143438677-143438677 on Build GRCh38
rs746385932 Chr.4: 143438678-143438678 on Build GRCh38

View Full Product Details