Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 48256507-48256778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LONP2
Entrez Gene ID
83752
Gene Name
lon peptidase 2, peroxisomal
Gene Aliases
LONP, LONPL
UniGene
Hs.295923
Gene Chromosome Location
Chr.16: 48244167-48353979 on Build GRCh38
Transcripts
NM_001300948.1 NM_031490.3 XM_017023755.1 XM_017023756.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6702119 c.488T>G p.M163R Chr.16: 48256629-48256629 on Build GRCh38
7078805 c.513G>A p.L171L Chr.16: 48256654-48256654 on Build GRCh38
1180040 c.471G>T p.L157F Chr.16: 48256612-48256612 on Build GRCh38
4438696 c.520C>T p.L174F Chr.16: 48256661-48256661 on Build GRCh38
3794875 c.591G>A p.E197E Chr.16: 48256732-48256732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867867730 Chr.16: 48256692-48256692 on Build GRCh38
rs576255071 Chr.16: 48256566-48256566 on Build GRCh38
rs762730600 Chr.16: 48256714-48256714 on Build GRCh38
rs759469371 Chr.16: 48256592-48256592 on Build GRCh38
rs193920895 Chr.16: 48256612-48256612 on Build GRCh38
rs905558859 Chr.16: 48256719-48256719 on Build GRCh38
rs755232608 Chr.16: 48256597-48256597 on Build GRCh38
rs779488956 Chr.16: 48256598-48256598 on Build GRCh38
rs929616882 Chr.16: 48256604-48256604 on Build GRCh38
rs763860627 Chr.16: 48256716-48256716 on Build GRCh38
rs748641719 Chr.16: 48256599-48256599 on Build GRCh38
rs750397946 Chr.16: 48256754-48256754 on Build GRCh38
rs772372264 Chr.16: 48256562-48256562 on Build GRCh38
rs527319828 Chr.16: 48256590-48256590 on Build GRCh38
rs756252897 Chr.16: 48256581-48256581 on Build GRCh38
rs774205883 Chr.16: 48256574-48256574 on Build GRCh38
rs774934578 Chr.16: 48256684-48256684 on Build GRCh38
rs774183905 Chr.16: 48256720-48256720 on Build GRCh38
rs756923617 Chr.16: 48256703-48256703 on Build GRCh38
rs749538944 Chr.16: 48256596-48256596 on Build GRCh38
rs200319783 Chr.16: 48256565-48256565 on Build GRCh38
rs935533442 Chr.16: 48256532-48256532 on Build GRCh38
rs773886549 Chr.16: 48256630-48256630 on Build GRCh38
rs771794983 Chr.16: 48256591-48256592 on Build GRCh38
rs541834463 Chr.16: 48256622-48256622 on Build GRCh38
rs745313902 Chr.16: 48256659-48256659 on Build GRCh38
rs1046800985 Chr.16: 48256652-48256652 on Build GRCh38
rs968367460 Chr.16: 48256537-48256537 on Build GRCh38
rs767406516 Chr.16: 48256753-48256753 on Build GRCh38
rs990108674 Chr.16: 48256547-48256547 on Build GRCh38
rs926816738 Chr.16: 48256555-48256555 on Build GRCh38
rs761712347 Chr.16: 48256728-48256728 on Build GRCh38
rs769356093 Chr.16: 48256662-48256662 on Build GRCh38

View Full Product Details