Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63244035-63244270 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKAIN4
Entrez Gene ID
128414
Gene Name
Na+/K+ transporting ATPase interacting 4
Gene Aliases
C20orf58, FAM77A, bA261N11.2
UniGene
Hs.46627
Gene Chromosome Location
Chr.20: 63240784-63254904 on Build GRCh38
Transcripts
NM_152864.3 XM_005260192.1 XM_005260194.3 XM_011528527.1 XM_011528528.2 XM_011528529.2 XM_017027636.1 XM_017027637.1 XM_017027638.1 XM_017027639.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2931513 c.507C>A p.S169R Chr.20: 63244049-63244049 on Build GRCh38
6231913 c.515C>T p.T172M Chr.20: 63244041-63244041 on Build GRCh38
368316 c.509T>G p.V170G Chr.20: 63244047-63244047 on Build GRCh38
1029036 c.508G>A p.V170M Chr.20: 63244048-63244048 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746646032 Chr.20: 63244127-63244127 on Build GRCh38
rs774456050 Chr.20: 63244109-63244109 on Build GRCh38
rs537794224 Chr.20: 63244116-63244116 on Build GRCh38
rs879621681 Chr.20: 63244205-63244205 on Build GRCh38
rs544804679 Chr.20: 63244146-63244146 on Build GRCh38
rs891807968 Chr.20: 63244181-63244181 on Build GRCh38
rs369757303 Chr.20: 63244098-63244098 on Build GRCh38
rs41282998 Chr.20: 63244161-63244161 on Build GRCh38
rs767537337 Chr.20: 63244113-63244113 on Build GRCh38
rs952636797 Chr.20: 63244184-63244184 on Build GRCh38
rs992617979 Chr.20: 63244237-63244237 on Build GRCh38
rs62207448 Chr.20: 63244187-63244187 on Build GRCh38
rs764383832 Chr.20: 63244117-63244117 on Build GRCh38
rs994591560 Chr.20: 63244148-63244148 on Build GRCh38
rs878968305 Chr.20: 63244149-63244149 on Build GRCh38
rs779571617 Chr.20: 63244126-63244126 on Build GRCh38
rs77939763 Chr.20: 63244144-63244144 on Build GRCh38
rs778268245 Chr.20: 63244109-63244110 on Build GRCh38
rs200603273 Chr.20: 63244108-63244108 on Build GRCh38
rs796931766 Chr.20: 63244109-63244110 on Build GRCh38
rs746622845 Chr.20: 63244064-63244064 on Build GRCh38
rs757907801 Chr.20: 63244121-63244121 on Build GRCh38
rs1047666693 Chr.20: 63244122-63244122 on Build GRCh38
rs2236186 Chr.20: 63244145-63244145 on Build GRCh38
rs754219420 Chr.20: 63244118-63244118 on Build GRCh38
rs761751287 Chr.20: 63244096-63244096 on Build GRCh38
rs958773356 Chr.20: 63244220-63244220 on Build GRCh38
rs542245629 Chr.20: 63244164-63244164 on Build GRCh38
rs779528124 Chr.20: 63244124-63244124 on Build GRCh38
rs752894141 Chr.20: 63244114-63244114 on Build GRCh38
rs776417374 Chr.20: 63244079-63244079 on Build GRCh38
rs759590975 Chr.20: 63244111-63244111 on Build GRCh38
rs927165455 Chr.20: 63244211-63244211 on Build GRCh38
rs976113825 Chr.20: 63244103-63244103 on Build GRCh38
rs559962254 Chr.20: 63244159-63244159 on Build GRCh38
rs768701334 Chr.20: 63244142-63244142 on Build GRCh38
rs758001381 Chr.20: 63244125-63244125 on Build GRCh38
rs367985350 Chr.20: 63244248-63244248 on Build GRCh38
rs768363146 Chr.20: 63244074-63244074 on Build GRCh38
rs1010892159 Chr.20: 63244232-63244232 on Build GRCh38

View Full Product Details