Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 169796650-169796919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC34
Entrez Gene ID
151827
Gene Name
leucine rich repeat containing 34
Gene Aliases
-
UniGene
Hs.591289
Gene Chromosome Location
Chr.3: 169793006-169812904 on Build GRCh38
Transcripts
NM_001172779.1 NM_001172780.1 NM_153353.4 XM_006713508.3 XM_011512442.2 XM_017005746.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3203841 c.644G>A p.R215H Chr.3: 169796874-169796874 on Build GRCh38
3915329 c.675A>G p.A225A Chr.3: 169796843-169796843 on Build GRCh38
6436733 c.819G>T p.M273I Chr.3: 169796834-169796834 on Build GRCh38
5658440 c.711T>C p.G237G Chr.3: 169796807-169796807 on Build GRCh38
5829632 c.856T>A p.L286I Chr.3: 169796797-169796797 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778622188 Chr.3: 169796875-169796875 on Build GRCh38
rs778348749 Chr.3: 169796799-169796800 on Build GRCh38
rs373594080 Chr.3: 169796852-169796852 on Build GRCh38
rs964987414 Chr.3: 169796784-169796784 on Build GRCh38
rs759210620 Chr.3: 169796800-169796800 on Build GRCh38
rs774004182 Chr.3: 169796783-169796783 on Build GRCh38
rs143470723 Chr.3: 169796763-169796763 on Build GRCh38
rs770495376 Chr.3: 169796849-169796849 on Build GRCh38
rs571979799 Chr.3: 169796874-169796874 on Build GRCh38
rs751418095 Chr.3: 169796730-169796736 on Build GRCh38
rs770985781 Chr.3: 169796854-169796855 on Build GRCh38
rs756073051 Chr.3: 169796839-169796839 on Build GRCh38
rs1020837801 Chr.3: 169796708-169796708 on Build GRCh38
rs112320485 Chr.3: 169796727-169796727 on Build GRCh38
rs765175081 Chr.3: 169796820-169796820 on Build GRCh38
rs781178804 Chr.3: 169796837-169796837 on Build GRCh38
rs373819156 Chr.3: 169796768-169796768 on Build GRCh38
rs372142875 Chr.3: 169796871-169796871 on Build GRCh38
rs759141342 Chr.3: 169796794-169796794 on Build GRCh38
rs754835580 Chr.3: 169796712-169796712 on Build GRCh38
rs547360001 Chr.3: 169796873-169796873 on Build GRCh38
rs777894364 Chr.3: 169796811-169796812 on Build GRCh38
rs749500302 Chr.3: 169796835-169796836 on Build GRCh38
rs750505809 Chr.3: 169796881-169796881 on Build GRCh38
rs775801012 Chr.3: 169796802-169796802 on Build GRCh38
rs142362395 Chr.3: 169796832-169796832 on Build GRCh38
rs530645050 Chr.3: 169796805-169796805 on Build GRCh38
rs777769698 Chr.3: 169796843-169796843 on Build GRCh38
rs748826240 Chr.3: 169796764-169796764 on Build GRCh38
rs750702166 Chr.3: 169796827-169796827 on Build GRCh38
rs766700734 Chr.3: 169796886-169796886 on Build GRCh38
rs780080945 Chr.3: 169796719-169796719 on Build GRCh38
rs533015571 Chr.3: 169796757-169796757 on Build GRCh38
rs769745350 Chr.3: 169796759-169796759 on Build GRCh38
rs867999463 Chr.3: 169796803-169796803 on Build GRCh38
rs10936600 Chr.3: 169796797-169796797 on Build GRCh38
rs146227830 Chr.3: 169796744-169796744 on Build GRCh38
rs747938538 Chr.3: 169796838-169796838 on Build GRCh38
rs781092428 Chr.3: 169796747-169796747 on Build GRCh38
rs950764607 Chr.3: 169796693-169796693 on Build GRCh38
rs745561009 Chr.3: 169796856-169796856 on Build GRCh38
rs146384166 Chr.3: 169796867-169796867 on Build GRCh38
rs764653260 Chr.3: 169796810-169796810 on Build GRCh38
rs184655709 Chr.3: 169796818-169796818 on Build GRCh38
rs201395437 Chr.3: 169796844-169796844 on Build GRCh38
rs771692142 Chr.3: 169796866-169796866 on Build GRCh38
rs751665358 Chr.3: 169796887-169796887 on Build GRCh38
rs376612149 Chr.3: 169796882-169796882 on Build GRCh38
rs765445391 Chr.3: 169796879-169796879 on Build GRCh38
rs753936489 Chr.3: 169796816-169796816 on Build GRCh38

View Full Product Details