Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 219979900-219980143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPRS
Entrez Gene ID
2058
Gene Name
glutamyl-prolyl-tRNA synthetase
Gene Aliases
EARS, GLUPRORS, PARS, PIG32, QARS, QPRS
UniGene
Hs.497788
Gene Chromosome Location
Chr.1: 219968600-220046658 on Build GRCh38
Transcripts
NM_004446.2 XM_017000614.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4028360 c.3671C>T p.T1224I Chr.1: 219980125-219980125 on Build GRCh38
1637868 c.3710A>T p.Q1237L Chr.1: 219980086-219980086 on Build GRCh38
5951466 c.3675A>C p.I1225I Chr.1: 219980121-219980121 on Build GRCh38
4681442 c.3697G>A p.G1233R Chr.1: 219980099-219980099 on Build GRCh38
1668487 c.3679G>T p.A1227S Chr.1: 219980117-219980117 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs3835619 Chr.1: 219979990-219979991 on Build GRCh38
rs919129762 Chr.1: 219979990-219979990 on Build GRCh38
rs868402451 Chr.1: 219980084-219980084 on Build GRCh38
rs768247332 Chr.1: 219980051-219980051 on Build GRCh38
rs775396857 Chr.1: 219980093-219980093 on Build GRCh38
rs117101042 Chr.1: 219980061-219980061 on Build GRCh38
rs774011588 Chr.1: 219980057-219980058 on Build GRCh38
rs759903052 Chr.1: 219980067-219980067 on Build GRCh38
rs778513654 Chr.1: 219980057-219980057 on Build GRCh38
rs747413517 Chr.1: 219980060-219980060 on Build GRCh38
rs78464168 Chr.1: 219979993-219979993 on Build GRCh38
rs926623662 Chr.1: 219980024-219980024 on Build GRCh38
rs559445163 Chr.1: 219979986-219979986 on Build GRCh38
rs963166400 Chr.1: 219979965-219979965 on Build GRCh38
rs985832281 Chr.1: 219979944-219979944 on Build GRCh38
rs950430230 Chr.1: 219980052-219980052 on Build GRCh38
rs749001631 Chr.1: 219980042-219980042 on Build GRCh38
rs34917248 Chr.1: 219979992-219979993 on Build GRCh38
rs763262450 Chr.1: 219980101-219980101 on Build GRCh38
rs997185880 Chr.1: 219980036-219980036 on Build GRCh38
rs386369689 Chr.1: 219979991-219979992 on Build GRCh38
rs1053444135 Chr.1: 219979950-219979950 on Build GRCh38
rs953187357 Chr.1: 219979928-219979928 on Build GRCh38
rs373651698 Chr.1: 219980037-219980037 on Build GRCh38
rs7537559 Chr.1: 219979964-219979964 on Build GRCh38
rs868611544 Chr.1: 219979996-219979996 on Build GRCh38
rs532166962 Chr.1: 219980005-219980005 on Build GRCh38
rs908773497 Chr.1: 219980062-219980062 on Build GRCh38
rs964514595 Chr.1: 219980006-219980006 on Build GRCh38
rs370984363 Chr.1: 219980081-219980081 on Build GRCh38

View Full Product Details