Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70674381-70674618 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTSS1L
Entrez Gene ID
92154
Gene Name
metastasis suppressor 1 like
Gene Aliases
ABBA-1, ABBA1
UniGene
Hs.432387
Gene Chromosome Location
Chr.16: 70661204-70686207 on Build GRCh38
Transcripts
NM_138383.2 XM_005256236.4 XM_011523450.2 XM_011523453.2 XM_011523454.2 XM_011523458.2 XM_017023879.1 XM_017023880.1 XM_017023881.1 XM_017023882.1 XM_017023883.1 XM_017023884.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5599961 c.971C>T p.S324F Chr.16: 70674388-70674388 on Build GRCh38
3273347 c.896A>G p.Y299C Chr.16: 70674463-70674463 on Build GRCh38
3273346 c.943A>G p.T315A Chr.16: 70674416-70674416 on Build GRCh38
6260842 c.934C>A p.Q312K Chr.16: 70674425-70674425 on Build GRCh38
4510401 c.836C>T p.P279L Chr.16: 70674523-70674523 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029144836 Chr.16: 70674599-70674599 on Build GRCh38
rs147072636 Chr.16: 70674436-70674436 on Build GRCh38
rs201486872 Chr.16: 70674525-70674525 on Build GRCh38
rs754624096 Chr.16: 70674566-70674566 on Build GRCh38
rs776111852 Chr.16: 70674540-70674540 on Build GRCh38
rs143838443 Chr.16: 70674490-70674490 on Build GRCh38
rs754505389 Chr.16: 70674424-70674424 on Build GRCh38
rs745862900 Chr.16: 70674448-70674448 on Build GRCh38
rs754384079 Chr.16: 70674422-70674422 on Build GRCh38
rs1001488030 Chr.16: 70674530-70674530 on Build GRCh38
rs747288873 Chr.16: 70674522-70674522 on Build GRCh38
rs1015022746 Chr.16: 70674562-70674562 on Build GRCh38
rs762334830 Chr.16: 70674544-70674544 on Build GRCh38
rs767787781 Chr.16: 70674410-70674410 on Build GRCh38
rs376758699 Chr.16: 70674443-70674443 on Build GRCh38
rs559783450 Chr.16: 70674432-70674432 on Build GRCh38
rs780410936 Chr.16: 70674407-70674408 on Build GRCh38
rs375933801 Chr.16: 70674416-70674416 on Build GRCh38
rs550061384 Chr.16: 70674407-70674407 on Build GRCh38
rs765777107 Chr.16: 70674480-70674480 on Build GRCh38
rs1001798972 Chr.16: 70674579-70674579 on Build GRCh38
rs757108365 Chr.16: 70674493-70674493 on Build GRCh38
rs1046148489 Chr.16: 70674546-70674546 on Build GRCh38
rs925819016 Chr.16: 70674438-70674438 on Build GRCh38
rs200193469 Chr.16: 70674489-70674489 on Build GRCh38
rs748292625 Chr.16: 70674463-70674463 on Build GRCh38
rs374576681 Chr.16: 70674468-70674468 on Build GRCh38
rs778796000 Chr.16: 70674453-70674453 on Build GRCh38
rs762544499 Chr.16: 70674484-70674484 on Build GRCh38
rs768688337 Chr.16: 70674406-70674406 on Build GRCh38
rs374842861 Chr.16: 70674533-70674533 on Build GRCh38
rs138255587 Chr.16: 70674437-70674437 on Build GRCh38
rs763758941 Chr.16: 70674551-70674551 on Build GRCh38
rs755988578 Chr.16: 70674578-70674578 on Build GRCh38
rs748156615 Chr.16: 70674510-70674510 on Build GRCh38
rs766848780 Chr.16: 70674419-70674419 on Build GRCh38
rs113024761 Chr.16: 70674433-70674433 on Build GRCh38
rs772179034 Chr.16: 70674514-70674514 on Build GRCh38
rs551681033 Chr.16: 70674598-70674598 on Build GRCh38
rs895819742 Chr.16: 70674541-70674541 on Build GRCh38
rs1042936577 Chr.16: 70674505-70674505 on Build GRCh38
rs781080424 Chr.16: 70674494-70674494 on Build GRCh38
rs757163615 Chr.16: 70674442-70674442 on Build GRCh38
rs751131819 Chr.16: 70674556-70674556 on Build GRCh38
rs142750803 Chr.16: 70674408-70674408 on Build GRCh38
rs750138628 Chr.16: 70674571-70674571 on Build GRCh38
rs780064481 Chr.16: 70674497-70674497 on Build GRCh38
rs760997239 Chr.16: 70674475-70674475 on Build GRCh38
rs769786397 Chr.16: 70674450-70674450 on Build GRCh38
rs147002775 Chr.16: 70674495-70674495 on Build GRCh38
rs776729070 Chr.16: 70674403-70674403 on Build GRCh38
rs1027810810 Chr.16: 70674592-70674592 on Build GRCh38
rs761052246 Chr.16: 70674415-70674415 on Build GRCh38
rs370244103 Chr.16: 70674481-70674481 on Build GRCh38
rs777109072 Chr.16: 70674476-70674476 on Build GRCh38
rs749353794 Chr.16: 70674404-70674404 on Build GRCh38
rs374908099 Chr.16: 70674519-70674519 on Build GRCh38
rs759975511 Chr.16: 70674527-70674527 on Build GRCh38
rs539780323 Chr.16: 70674426-70674426 on Build GRCh38
rs372416793 Chr.16: 70674559-70674559 on Build GRCh38
rs547591161 Chr.16: 70674427-70674427 on Build GRCh38
rs371325152 Chr.16: 70674465-70674465 on Build GRCh38
rs551572143 Chr.16: 70674567-70674567 on Build GRCh38
rs200666822 Chr.16: 70674478-70674478 on Build GRCh38
rs751273440 Chr.16: 70674422-70674424 on Build GRCh38
rs773526510 Chr.16: 70674412-70674412 on Build GRCh38

View Full Product Details