Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 38367228-38367499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTC
Entrez Gene ID
5009
Gene Name
ornithine carbamoyltransferase
Gene Aliases
OCTD
UniGene
Hs.117050
Gene Chromosome Location
Chr.X: 38352483-38421450 on Build GRCh38
Transcripts
NM_000531.5 XM_017029556.1

SNP Detail

SNP ID Chromosome Location
rs753723228 Chr.X: 38367444-38367444 on Build GRCh38
rs763821262 Chr.X: 38367424-38367424 on Build GRCh38
rs199858968 Chr.X: 38367296-38367296 on Build GRCh38
rs72554322 Chr.X: 38367387-38367387 on Build GRCh38
rs72554313 Chr.X: 38367348-38367348 on Build GRCh38
rs72554305 Chr.X: 38367319-38367319 on Build GRCh38
rs72554324 Chr.X: 38367401-38367401 on Build GRCh38
rs72554308 Chr.X: 38367332-38367332 on Build GRCh38
rs774764719 Chr.X: 38367438-38367438 on Build GRCh38
rs763827802 Chr.X: 38367469-38367469 on Build GRCh38
rs775711263 Chr.X: 38367451-38367451 on Build GRCh38
rs72554310 Chr.X: 38367344-38367344 on Build GRCh38
rs748525448 Chr.X: 38367259-38367259 on Build GRCh38
rs72554317 Chr.X: 38367368-38367368 on Build GRCh38
rs759639432 Chr.X: 38367293-38367293 on Build GRCh38
rs753631467 Chr.X: 38367428-38367428 on Build GRCh38
rs72554320 Chr.X: 38367380-38367380 on Build GRCh38
rs766547928 Chr.X: 38367473-38367473 on Build GRCh38
rs74518351 Chr.X: 38367335-38367335 on Build GRCh38
rs375148070 Chr.X: 38367452-38367452 on Build GRCh38
rs66677059 Chr.X: 38367371-38367371 on Build GRCh38
rs72554307 Chr.X: 38367331-38367331 on Build GRCh38
rs141273695 Chr.X: 38367291-38367291 on Build GRCh38
rs763877023 Chr.X: 38367301-38367301 on Build GRCh38
rs72554326 Chr.X: 38367418-38367418 on Build GRCh38
rs924740805 Chr.X: 38367384-38367384 on Build GRCh38
rs907001683 Chr.X: 38367472-38367473 on Build GRCh38
rs766512759 Chr.X: 38367286-38367286 on Build GRCh38
rs72554323 Chr.X: 38367392-38367392 on Build GRCh38
rs772107914 Chr.X: 38367448-38367448 on Build GRCh38
rs67939655 Chr.X: 38367353-38367353 on Build GRCh38
rs72554316 Chr.X: 38367358-38367358 on Build GRCh38
rs993705802 Chr.X: 38367294-38367294 on Build GRCh38
rs935161024 Chr.X: 38367385-38367385 on Build GRCh38
rs760784385 Chr.X: 38367442-38367443 on Build GRCh38
rs72554303 Chr.X: 38367288-38367288 on Build GRCh38
rs72554315 Chr.X: 38367356-38367356 on Build GRCh38
rs886041526 Chr.X: 38367430-38367430 on Build GRCh38
rs72554319 Chr.X: 38367376-38367376 on Build GRCh38
rs72554312 Chr.X: 38367347-38367347 on Build GRCh38
rs201802621 Chr.X: 38367362-38367362 on Build GRCh38
rs987967261 Chr.X: 38367429-38367429 on Build GRCh38
rs144153859 Chr.X: 38367360-38367360 on Build GRCh38
rs752916728 Chr.X: 38367298-38367298 on Build GRCh38
rs72554304 Chr.X: 38367307-38367307 on Build GRCh38
rs761634797 Chr.X: 38367476-38367476 on Build GRCh38
rs369756595 Chr.X: 38367409-38367409 on Build GRCh38
rs369331778 Chr.X: 38367473-38367473 on Build GRCh38
rs72554306 Chr.X: 38367328-38367328 on Build GRCh38
rs750152681 Chr.X: 38367260-38367260 on Build GRCh38
rs72554321 Chr.X: 38367383-38367383 on Build GRCh38
rs72554318 Chr.X: 38367369-38367369 on Build GRCh38
rs67486158 Chr.X: 38367361-38367361 on Build GRCh38
rs66693137 Chr.X: 38367430-38367430 on Build GRCh38
rs66521141 Chr.X: 38367367-38367367 on Build GRCh38
rs765197868 Chr.X: 38367477-38367477 on Build GRCh38
rs750386308 Chr.X: 38367478-38367478 on Build GRCh38
rs796052011 Chr.X: 38367277-38367277 on Build GRCh38
rs763559328 Chr.X: 38367268-38367268 on Build GRCh38
rs72554309 Chr.X: 38367340-38367340 on Build GRCh38
rs759645083 Chr.X: 38367447-38367447 on Build GRCh38
rs72554325 Chr.X: 38367413-38367413 on Build GRCh38
rs1800321 Chr.X: 38367350-38367350 on Build GRCh38
rs753712951 Chr.X: 38367302-38367302 on Build GRCh38
rs72554311 Chr.X: 38367346-38367346 on Build GRCh38
rs757207910 Chr.X: 38367318-38367318 on Build GRCh38
rs759104592 Chr.X: 38367299-38367299 on Build GRCh38
rs72554314 Chr.X: 38367353-38367354 on Build GRCh38
rs281865554 Chr.X: 38367290-38367290 on Build GRCh38

View Full Product Details