Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 50152800-50153071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAPD5
Entrez Gene ID
64282
Gene Name
PAP associated domain containing 5
Gene Aliases
TRF4-2
UniGene
Hs.514342
Gene Chromosome Location
Chr.16: 50152918-50235310 on Build GRCh38
Transcripts
NM_001040284.2 NM_001040285.2 XM_005256093.4 XM_011523275.2 XM_011523276.2 XM_011523278.2 XM_017023555.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1609379 c.14C>A p.P5Q Chr.16: 50152966-50152966 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs898904566 Chr.16: 50152852-50152852 on Build GRCh38
rs916380972 Chr.16: 50152970-50152970 on Build GRCh38
rs549071389 Chr.16: 50152859-50152859 on Build GRCh38
rs965349917 Chr.16: 50152939-50152939 on Build GRCh38
rs563760022 Chr.16: 50152844-50152844 on Build GRCh38
rs990005331 Chr.16: 50152959-50152959 on Build GRCh38
rs962770605 Chr.16: 50152982-50152995 on Build GRCh38
rs1007489015 Chr.16: 50153047-50153047 on Build GRCh38
rs772372381 Chr.16: 50153052-50153052 on Build GRCh38
rs551350671 Chr.16: 50152887-50152887 on Build GRCh38
rs898867858 Chr.16: 50153033-50153033 on Build GRCh38
rs995021995 Chr.16: 50152901-50152901 on Build GRCh38
rs559704335 Chr.16: 50152946-50152946 on Build GRCh38
rs1029168515 Chr.16: 50153042-50153042 on Build GRCh38
rs936766959 Chr.16: 50152954-50152954 on Build GRCh38
rs985698244 Chr.16: 50152935-50152935 on Build GRCh38
rs978100600 Chr.16: 50152945-50152945 on Build GRCh38
rs1019881665 Chr.16: 50152937-50152937 on Build GRCh38
rs889923881 Chr.16: 50153046-50153046 on Build GRCh38
rs750465908 Chr.16: 50152923-50152923 on Build GRCh38
rs539232823 Chr.16: 50153014-50153014 on Build GRCh38
rs778986759 Chr.16: 50153002-50153002 on Build GRCh38
rs1046274249 Chr.16: 50152980-50152980 on Build GRCh38
rs529407309 Chr.16: 50152845-50152845 on Build GRCh38
rs527544581 Chr.16: 50152951-50152951 on Build GRCh38
rs570453581 Chr.16: 50152979-50152979 on Build GRCh38
rs940639022 Chr.16: 50152990-50152990 on Build GRCh38
rs1027154104 Chr.16: 50152932-50152932 on Build GRCh38
rs755943174 Chr.16: 50152928-50152928 on Build GRCh38
rs547285140 Chr.16: 50152956-50152956 on Build GRCh38
rs994590772 Chr.16: 50153039-50153039 on Build GRCh38

View Full Product Details