Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 152736606-152736371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSAG4
Entrez Gene ID
100130935
Gene Name
CSAG family member 4 (pseudogene)
Gene Aliases
-
UniGene
Hs.169246
Gene Chromosome Location
Chr.X: 152733827-152740985 on Build GRCh38
Transcripts
NR_073432.1 NM_001166386.3 NM_001166387.3 NM_005367.6

Gene Symbol
MAGEA12
Entrez Gene ID
4111
Gene Name
MAGE family member A12
Gene Aliases
CT1.12, MAGE12
UniGene
Hs.169246
Gene Chromosome Location
Chr.X: 152733779-152737669 on Build GRCh38
Transcripts
NR_073432.1 NM_001166386.3 NM_001166387.3 NM_005367.6

View Full Product Details