Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 115525246-115525507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEM1C
Entrez Gene ID
56929
Gene Name
fem-1 homolog C
Gene Aliases
EUROIMAGE686608, EUROIMAGE783647, FEM1A
UniGene
Hs.47367
Gene Chromosome Location
Chr.5: 115520908-115544919 on Build GRCh38
Transcripts
NM_020177.2 XM_005272035.4 XM_017009647.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6447100 c.783T>A p.D261E Chr.5: 115525379-115525379 on Build GRCh38
1252352 c.778_779insA p.R260fs*20 Chr.5: 115525383-115525384 on Build GRCh38
6584525 c.916G>C p.E306Q Chr.5: 115525246-115525246 on Build GRCh38
7368115 c.706C>T p.H236Y Chr.5: 115525456-115525456 on Build GRCh38
6239817 c.777A>C p.K259N Chr.5: 115525385-115525385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775053257 Chr.5: 115525342-115525342 on Build GRCh38
rs147529913 Chr.5: 115525440-115525440 on Build GRCh38
rs768115858 Chr.5: 115525376-115525376 on Build GRCh38
rs529545977 Chr.5: 115525345-115525345 on Build GRCh38
rs923259011 Chr.5: 115525420-115525420 on Build GRCh38
rs140001534 Chr.5: 115525286-115525286 on Build GRCh38
rs144957300 Chr.5: 115525332-115525332 on Build GRCh38
rs757782894 Chr.5: 115525294-115525294 on Build GRCh38
rs781708327 Chr.5: 115525304-115525304 on Build GRCh38
rs752932852 Chr.5: 115525477-115525477 on Build GRCh38
rs761749339 Chr.5: 115525396-115525396 on Build GRCh38
rs567959573 Chr.5: 115525438-115525438 on Build GRCh38
rs548208267 Chr.5: 115525431-115525431 on Build GRCh38
rs758468709 Chr.5: 115525285-115525285 on Build GRCh38
rs770164336 Chr.5: 115525334-115525334 on Build GRCh38
rs758582155 Chr.5: 115525479-115525479 on Build GRCh38
rs773781743 Chr.5: 115525379-115525379 on Build GRCh38
rs976333369 Chr.5: 115525338-115525338 on Build GRCh38
rs375159711 Chr.5: 115525432-115525432 on Build GRCh38
rs752945818 Chr.5: 115525274-115525274 on Build GRCh38
rs965302978 Chr.5: 115525292-115525292 on Build GRCh38
rs764284851 Chr.5: 115525487-115525487 on Build GRCh38
rs765838006 Chr.5: 115525433-115525433 on Build GRCh38
rs749480301 Chr.5: 115525289-115525289 on Build GRCh38

View Full Product Details