Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112231727-112231986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN7
Entrez Gene ID
129804
Gene Name
fibulin 7
Gene Aliases
TM14
UniGene
Hs.437696
Gene Chromosome Location
Chr.2: 112138385-112254888 on Build GRCh38
Transcripts
XM_017003317.1 NM_032494.2 XM_011511998.1 XM_017005107.1

Gene Symbol
ZC3H8
Entrez Gene ID
84524
Gene Name
zinc finger CCCH-type containing 8
Gene Aliases
Fliz1, ZC3HDC8
UniGene
Hs.418416
Gene Chromosome Location
Chr.2: 112215035-112255087 on Build GRCh38
Transcripts
XM_017003317.1 NM_032494.2 XM_011511998.1 XM_017005107.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4922442 c.748A>C p.K250Q Chr.2: 112231933-112231933 on Build GRCh38
7448809 c.839C>A p.A280D Chr.2: 112231842-112231842 on Build GRCh38
1005498 c.798T>G p.F266L Chr.2: 112231883-112231883 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs563261710 Chr.2: 112231807-112231807 on Build GRCh38
rs748316181 Chr.2: 112231879-112231879 on Build GRCh38
rs774812862 Chr.2: 112231929-112231929 on Build GRCh38
rs770061992 Chr.2: 112231884-112231884 on Build GRCh38
rs766836197 Chr.2: 112231912-112231912 on Build GRCh38
rs778565181 Chr.2: 112231819-112231819 on Build GRCh38
rs778735347 Chr.2: 112231811-112231811 on Build GRCh38
rs945375235 Chr.2: 112231823-112231823 on Build GRCh38
rs776830085 Chr.2: 112231868-112231868 on Build GRCh38
rs779614609 Chr.2: 112231804-112231805 on Build GRCh38
rs376119163 Chr.2: 112231804-112231804 on Build GRCh38
rs777271418 Chr.2: 112231854-112231855 on Build GRCh38
rs781135871 Chr.2: 112231952-112231952 on Build GRCh38
rs936966588 Chr.2: 112231755-112231755 on Build GRCh38
rs773511770 Chr.2: 112231895-112231895 on Build GRCh38
rs768598046 Chr.2: 112231864-112231864 on Build GRCh38
rs768341644 Chr.2: 112231838-112231838 on Build GRCh38
rs763387471 Chr.2: 112231789-112231789 on Build GRCh38
rs773646250 Chr.2: 112231754-112231757 on Build GRCh38
rs915960186 Chr.2: 112231945-112231945 on Build GRCh38
rs756989772 Chr.2: 112231814-112231814 on Build GRCh38
rs576887974 Chr.2: 112231809-112231809 on Build GRCh38
rs746507987 Chr.2: 112231831-112231834 on Build GRCh38
rs867354910 Chr.2: 112231890-112231890 on Build GRCh38
rs763332483 Chr.2: 112231907-112231907 on Build GRCh38
rs780162705 Chr.2: 112231843-112231843 on Build GRCh38
rs758351113 Chr.2: 112231833-112231833 on Build GRCh38
rs747045212 Chr.2: 112231860-112231860 on Build GRCh38
rs745660635 Chr.2: 112231821-112231821 on Build GRCh38
rs1054186506 Chr.2: 112231767-112231767 on Build GRCh38
rs957510369 Chr.2: 112231760-112231760 on Build GRCh38

View Full Product Details