Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31625461-31625686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRRC2A
Entrez Gene ID
7916
Gene Name
proline rich coiled-coil 2A
Gene Aliases
BAT2, D6S51, D6S51E, G2
UniGene
Hs.123239
Gene Chromosome Location
Chr.6: 31620673-31637780 on Build GRCh38
Transcripts
NM_004638.3 NM_080686.2 XM_017011274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
317043 c.742delG p.G248fs*2 Chr.6: 31625594-31625594 on Build GRCh38
3861718 c.662C>A p.P221Q Chr.6: 31625514-31625514 on Build GRCh38
4414477 c.695G>A p.R232Q Chr.6: 31625547-31625547 on Build GRCh38
218874 c.638G>A p.R213H Chr.6: 31625490-31625490 on Build GRCh38
1077607 c.720A>C p.P240P Chr.6: 31625572-31625572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767729768 Chr.6: 31625645-31625645 on Build GRCh38
rs778124334 Chr.6: 31625513-31625513 on Build GRCh38
rs779373727 Chr.6: 31625501-31625501 on Build GRCh38
rs200705973 Chr.6: 31625572-31625572 on Build GRCh38
rs145609055 Chr.6: 31625602-31625602 on Build GRCh38
rs760065944 Chr.6: 31625553-31625553 on Build GRCh38
rs371703102 Chr.6: 31625530-31625530 on Build GRCh38
rs111897850 Chr.6: 31625612-31625612 on Build GRCh38
rs116604773 Chr.6: 31625547-31625547 on Build GRCh38
rs758486902 Chr.6: 31625506-31625506 on Build GRCh38
rs371386291 Chr.6: 31625619-31625619 on Build GRCh38
rs17200837 Chr.6: 31625488-31625488 on Build GRCh38
rs781198716 Chr.6: 31625586-31625586 on Build GRCh38
rs374903725 Chr.6: 31625539-31625539 on Build GRCh38
rs755359922 Chr.6: 31625493-31625493 on Build GRCh38
rs770881443 Chr.6: 31625536-31625541 on Build GRCh38
rs768601440 Chr.6: 31625534-31625534 on Build GRCh38
rs760069882 Chr.6: 31625630-31625632 on Build GRCh38
rs745958478 Chr.6: 31625591-31625591 on Build GRCh38
rs771615196 Chr.6: 31625544-31625544 on Build GRCh38
rs773958687 Chr.6: 31625538-31625538 on Build GRCh38
rs146068659 Chr.6: 31625515-31625515 on Build GRCh38
rs910207949 Chr.6: 31625637-31625637 on Build GRCh38
rs754443109 Chr.6: 31625490-31625490 on Build GRCh38
rs772705692 Chr.6: 31625607-31625607 on Build GRCh38
rs148710175 Chr.6: 31625543-31625543 on Build GRCh38
rs139649824 Chr.6: 31625503-31625503 on Build GRCh38
rs142466078 Chr.6: 31625546-31625546 on Build GRCh38
rs764027729 Chr.6: 31625605-31625605 on Build GRCh38
rs758855919 Chr.6: 31625574-31625574 on Build GRCh38
rs765648932 Chr.6: 31625632-31625632 on Build GRCh38
rs371020699 Chr.6: 31625639-31625639 on Build GRCh38
rs749181456 Chr.6: 31625533-31625533 on Build GRCh38
rs751942027 Chr.6: 31625640-31625640 on Build GRCh38
rs753424384 Chr.6: 31625551-31625551 on Build GRCh38
rs779936841 Chr.6: 31625528-31625528 on Build GRCh38
rs766024389 Chr.6: 31625548-31625548 on Build GRCh38
rs151316119 Chr.6: 31625578-31625578 on Build GRCh38
rs757503902 Chr.6: 31625583-31625583 on Build GRCh38
rs771050325 Chr.6: 31625521-31625521 on Build GRCh38
rs917109024 Chr.6: 31625518-31625518 on Build GRCh38
rs144276631 Chr.6: 31625514-31625514 on Build GRCh38
rs759187752 Chr.6: 31625626-31625626 on Build GRCh38
rs748468116 Chr.6: 31625624-31625624 on Build GRCh38
rs779190418 Chr.6: 31625644-31625644 on Build GRCh38
rs747878143 Chr.6: 31625604-31625604 on Build GRCh38
rs377026209 Chr.6: 31625611-31625611 on Build GRCh38
rs114452424 Chr.6: 31625592-31625592 on Build GRCh38
rs763197974 Chr.6: 31625635-31625635 on Build GRCh38
rs750498358 Chr.6: 31625648-31625648 on Build GRCh38
rs776895459 Chr.6: 31625623-31625627 on Build GRCh38
rs894020675 Chr.6: 31625631-31625631 on Build GRCh38

View Full Product Details