Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5997063-5997294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100128568
Entrez Gene ID
100128568
Gene Name
uncharacterized LOC100128568
Gene Aliases
-
UniGene
Hs.624525
Gene Chromosome Location
Chr.19: 5978403-6020363 on Build GRCh38
Transcripts
NR_046376.1 NM_000635.3 NM_134433.2 XM_011528171.2 XM_011528172.1 XM_017027107.1 XM_017027108.1

Gene Symbol
RFX2
Entrez Gene ID
5990
Gene Name
regulatory factor X2
Gene Aliases
-
UniGene
Hs.465709
Gene Chromosome Location
Chr.19: 5993164-6110653 on Build GRCh38
Transcripts
NR_046376.1 NM_000635.3 NM_134433.2 XM_011528171.2 XM_011528172.1 XM_017027107.1 XM_017027108.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6765360 c.1936G>A p.D646N Chr.19: 5997137-5997137 on Build GRCh38
3540839 c.1862C>T p.S621F Chr.19: 5997211-5997211 on Build GRCh38
6086206 c.1905C>T p.F635F Chr.19: 5997168-5997168 on Build GRCh38
5461013 c.1889G>A p.R630H Chr.19: 5997184-5997184 on Build GRCh38
3540838 c.1933T>C p.Y645H Chr.19: 5997140-5997140 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770125931 Chr.19: 5997107-5997107 on Build GRCh38
rs748517008 Chr.19: 5997230-5997230 on Build GRCh38
rs1015514275 Chr.19: 5997104-5997104 on Build GRCh38
rs773633075 Chr.19: 5997117-5997117 on Build GRCh38
rs75499120 Chr.19: 5997105-5997105 on Build GRCh38
rs778116547 Chr.19: 5997238-5997238 on Build GRCh38
rs757826142 Chr.19: 5997208-5997208 on Build GRCh38
rs202100959 Chr.19: 5997257-5997257 on Build GRCh38
rs555826981 Chr.19: 5997263-5997263 on Build GRCh38
rs371782022 Chr.19: 5997138-5997138 on Build GRCh38
rs138803994 Chr.19: 5997159-5997159 on Build GRCh38
rs781066965 Chr.19: 5997082-5997082 on Build GRCh38
rs762907288 Chr.19: 5997119-5997119 on Build GRCh38
rs182213065 Chr.19: 5997264-5997264 on Build GRCh38
rs764026497 Chr.19: 5997153-5997153 on Build GRCh38
rs764867079 Chr.19: 5997180-5997180 on Build GRCh38
rs139431127 Chr.19: 5997081-5997081 on Build GRCh38
rs753667887 Chr.19: 5997089-5997089 on Build GRCh38
rs541592870 Chr.19: 5997266-5997266 on Build GRCh38
rs372613961 Chr.19: 5997271-5997271 on Build GRCh38
rs779567495 Chr.19: 5997212-5997212 on Build GRCh38
rs745371197 Chr.19: 5997096-5997096 on Build GRCh38
rs757994512 Chr.19: 5997098-5997098 on Build GRCh38
rs199954102 Chr.19: 5997226-5997226 on Build GRCh38
rs200898749 Chr.19: 5997102-5997102 on Build GRCh38
rs149610449 Chr.19: 5997099-5997099 on Build GRCh38
rs576493331 Chr.19: 5997129-5997129 on Build GRCh38
rs774312384 Chr.19: 5997135-5997135 on Build GRCh38
rs751035286 Chr.19: 5997222-5997222 on Build GRCh38
rs201839547 Chr.19: 5997090-5997090 on Build GRCh38
rs912238139 Chr.19: 5997192-5997192 on Build GRCh38
rs12983826 Chr.19: 5997244-5997244 on Build GRCh38
rs760625463 Chr.19: 5997148-5997148 on Build GRCh38
rs571540930 Chr.19: 5997106-5997106 on Build GRCh38
rs147306532 Chr.19: 5997168-5997168 on Build GRCh38
rs750132162 Chr.19: 5997186-5997186 on Build GRCh38
rs370504934 Chr.19: 5997177-5997177 on Build GRCh38
rs375437676 Chr.19: 5997247-5997247 on Build GRCh38
rs766526459 Chr.19: 5997126-5997126 on Build GRCh38
rs189991674 Chr.19: 5997225-5997225 on Build GRCh38
rs770320295 Chr.19: 5997231-5997231 on Build GRCh38

View Full Product Details