Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 89037884-89038153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHC3
Entrez Gene ID
53358
Gene Name
SHC adaptor protein 3
Gene Aliases
N-Shc, NSHC, RAI, SHCC
UniGene
Hs.292737
Gene Chromosome Location
Chr.9: 89005771-89182308 on Build GRCh38
Transcripts
NM_016848.5 XM_011518785.2 XM_011518786.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3908748 c.218C>A p.T73N Chr.9: 89038075-89038075 on Build GRCh38
7209197 c.1602G>A p.T534T Chr.9: 89038047-89038047 on Build GRCh38
3908747 c.1574C>A p.T525N Chr.9: 89038075-89038075 on Build GRCh38
3774391 c.199T>C p.F67L Chr.9: 89038094-89038094 on Build GRCh38
3908746 c.245C>T p.T82M Chr.9: 89038048-89038048 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2282124 Chr.9: 89038110-89038110 on Build GRCh38
rs371151904 Chr.9: 89037981-89037981 on Build GRCh38
rs372829968 Chr.9: 89037967-89037967 on Build GRCh38
rs1007631191 Chr.9: 89038107-89038107 on Build GRCh38
rs62638717 Chr.9: 89038065-89038065 on Build GRCh38
rs760328967 Chr.9: 89037986-89037986 on Build GRCh38
rs56005403 Chr.9: 89038062-89038062 on Build GRCh38
rs868525357 Chr.9: 89037912-89037912 on Build GRCh38
rs1054566528 Chr.9: 89038080-89038080 on Build GRCh38
rs777317063 Chr.9: 89037964-89037964 on Build GRCh38
rs572901403 Chr.9: 89037961-89037961 on Build GRCh38
rs773640499 Chr.9: 89037962-89037963 on Build GRCh38
rs200227289 Chr.9: 89037965-89037965 on Build GRCh38
rs867373491 Chr.9: 89038092-89038092 on Build GRCh38
rs770677841 Chr.9: 89038042-89038042 on Build GRCh38
rs143304407 Chr.9: 89038038-89038038 on Build GRCh38
rs202188394 Chr.9: 89037979-89037979 on Build GRCh38
rs758951230 Chr.9: 89038013-89038013 on Build GRCh38
rs780414973 Chr.9: 89038015-89038015 on Build GRCh38
rs750898497 Chr.9: 89038011-89038011 on Build GRCh38
rs763743941 Chr.9: 89038056-89038056 on Build GRCh38
rs756456873 Chr.9: 89038100-89038100 on Build GRCh38
rs181529544 Chr.9: 89038094-89038094 on Build GRCh38
rs370776639 Chr.9: 89037993-89037993 on Build GRCh38
rs989711593 Chr.9: 89037954-89037954 on Build GRCh38
rs147547559 Chr.9: 89038045-89038045 on Build GRCh38
rs768780156 Chr.9: 89037971-89037971 on Build GRCh38
rs371185571 Chr.9: 89037985-89037985 on Build GRCh38
rs575391468 Chr.9: 89038047-89038047 on Build GRCh38
rs148359150 Chr.9: 89038017-89038017 on Build GRCh38
rs779397391 Chr.9: 89037966-89037966 on Build GRCh38
rs753323815 Chr.9: 89037988-89037988 on Build GRCh38
rs770929587 Chr.9: 89037977-89037977 on Build GRCh38
rs201006585 Chr.9: 89037974-89037974 on Build GRCh38
rs188886677 Chr.9: 89038059-89038059 on Build GRCh38
rs200275295 Chr.9: 89038040-89038040 on Build GRCh38
rs781279811 Chr.9: 89038035-89038035 on Build GRCh38
rs764795129 Chr.9: 89037994-89037994 on Build GRCh38
rs202001514 Chr.9: 89037944-89037944 on Build GRCh38
rs769994093 Chr.9: 89038039-89038039 on Build GRCh38
rs751889720 Chr.9: 89038074-89038074 on Build GRCh38
rs79508174 Chr.9: 89038108-89038108 on Build GRCh38
rs150513468 Chr.9: 89038101-89038101 on Build GRCh38
rs772159846 Chr.9: 89038044-89038044 on Build GRCh38
rs144984717 Chr.9: 89038066-89038066 on Build GRCh38
rs376350694 Chr.9: 89037968-89037968 on Build GRCh38
rs748353386 Chr.9: 89038097-89038097 on Build GRCh38
rs761799499 Chr.9: 89037975-89037975 on Build GRCh38
rs755281337 Chr.9: 89038088-89038088 on Build GRCh38
rs775304869 Chr.9: 89038119-89038119 on Build GRCh38

View Full Product Details