Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 130440859-130441084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM200A
Entrez Gene ID
114801
Gene Name
transmembrane protein 200A
Gene Aliases
KIAA1913, TTMA, TTMC
UniGene
Hs.591341
Gene Chromosome Location
Chr.6: 130366281-130443424 on Build GRCh38
Transcripts
NM_001258276.1 NM_001258277.1 NM_001258278.1 NM_052913.2 XM_005266816.2 XM_011535419.2 XM_017010230.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1440449 c.577G>A p.E193K Chr.6: 130440999-130440999 on Build GRCh38
7093666 c.611G>T p.R204I Chr.6: 130441033-130441033 on Build GRCh38
3620567 c.608C>T p.S203L Chr.6: 130441030-130441030 on Build GRCh38
6105207 c.642G>T p.S214S Chr.6: 130441064-130441064 on Build GRCh38
1544977 c.649C>T p.R217W Chr.6: 130441071-130441071 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763572984 Chr.6: 130440899-130440899 on Build GRCh38
rs551243409 Chr.6: 130441031-130441031 on Build GRCh38
rs986752652 Chr.6: 130440980-130440980 on Build GRCh38
rs776562446 Chr.6: 130441023-130441023 on Build GRCh38
rs761390006 Chr.6: 130440919-130440919 on Build GRCh38
rs767975924 Chr.6: 130441035-130441035 on Build GRCh38
rs13215027 Chr.6: 130441053-130441053 on Build GRCh38
rs149308469 Chr.6: 130440937-130440937 on Build GRCh38
rs145944902 Chr.6: 130441048-130441048 on Build GRCh38
rs755255101 Chr.6: 130440939-130440939 on Build GRCh38
rs925306035 Chr.6: 130440891-130440891 on Build GRCh38
rs768513368 Chr.6: 130441012-130441012 on Build GRCh38
rs746134252 Chr.6: 130440978-130440978 on Build GRCh38
rs756747178 Chr.6: 130440953-130440953 on Build GRCh38
rs201963998 Chr.6: 130441063-130441063 on Build GRCh38
rs777999253 Chr.6: 130440962-130440962 on Build GRCh38
rs775314072 Chr.6: 130440997-130440997 on Build GRCh38
rs931103745 Chr.6: 130440970-130440970 on Build GRCh38
rs767221082 Chr.6: 130440973-130440973 on Build GRCh38
rs757246248 Chr.6: 130441051-130441051 on Build GRCh38
rs13200367 Chr.6: 130440983-130440983 on Build GRCh38
rs778955775 Chr.6: 130440974-130440974 on Build GRCh38
rs1041082823 Chr.6: 130441060-130441060 on Build GRCh38
rs779091090 Chr.6: 130441052-130441052 on Build GRCh38
rs764831715 Chr.6: 130441030-130441030 on Build GRCh38
rs753412772 Chr.6: 130440911-130440911 on Build GRCh38
rs753229064 Chr.6: 130440950-130440950 on Build GRCh38
rs13215023 Chr.6: 130441033-130441033 on Build GRCh38
rs868348179 Chr.6: 130440991-130440991 on Build GRCh38
rs141608705 Chr.6: 130440987-130440987 on Build GRCh38
rs761177929 Chr.6: 130441028-130441028 on Build GRCh38
rs139970487 Chr.6: 130440968-130440968 on Build GRCh38
rs746944218 Chr.6: 130441003-130441003 on Build GRCh38
rs143602233 Chr.6: 130440938-130440938 on Build GRCh38
rs13214707 Chr.6: 130441034-130441034 on Build GRCh38
rs959386402 Chr.6: 130440895-130440895 on Build GRCh38
rs13214573 Chr.6: 130440944-130440944 on Build GRCh38
rs540895073 Chr.6: 130440969-130440969 on Build GRCh38
rs376673544 Chr.6: 130440898-130440898 on Build GRCh38
rs183247605 Chr.6: 130441049-130441049 on Build GRCh38
rs13200355 Chr.6: 130440946-130440946 on Build GRCh38
rs200933561 Chr.6: 130440943-130440943 on Build GRCh38

View Full Product Details