Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10106196-10106421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPAN
Entrez Gene ID
56342
Gene Name
peter pan homolog (Drosophila)
Gene Aliases
BXDC3, SSF, SSF-1, SSF1, SSF2
UniGene
Hs.14468
Gene Chromosome Location
Chr.19: 10106223-10111299 on Build GRCh38
Transcripts
NM_020230.5 NM_001040664.2 NM_001198690.1

Gene Symbol
PPAN-P2RY11
Entrez Gene ID
692312
Gene Name
PPAN-P2RY11 readthrough
Gene Aliases
-
UniGene
Hs.14468
Gene Chromosome Location
Chr.19: 10106223-10115389 on Build GRCh38
Transcripts
NM_020230.5 NM_001040664.2 NM_001198690.1

SNP Detail

SNP ID Chromosome Location
rs891472876 Chr.19: 10106271-10106271 on Build GRCh38
rs961377278 Chr.19: 10106366-10106366 on Build GRCh38
rs533306747 Chr.19: 10106240-10106240 on Build GRCh38
rs567384448 Chr.19: 10106303-10106303 on Build GRCh38
rs1053014298 Chr.19: 10106262-10106262 on Build GRCh38
rs993584064 Chr.19: 10106253-10106254 on Build GRCh38
rs867448059 Chr.19: 10106299-10106299 on Build GRCh38
rs2290685 Chr.19: 10106295-10106295 on Build GRCh38
rs748481680 Chr.19: 10106389-10106389 on Build GRCh38
rs979196160 Chr.19: 10106308-10106308 on Build GRCh38
rs766612887 Chr.19: 10106310-10106310 on Build GRCh38
rs970189463 Chr.19: 10106392-10106392 on Build GRCh38
rs556089220 Chr.19: 10106344-10106344 on Build GRCh38
rs756229840 Chr.19: 10106309-10106309 on Build GRCh38
rs566696390 Chr.19: 10106259-10106259 on Build GRCh38
rs925061860 Chr.19: 10106319-10106319 on Build GRCh38
rs758614227 Chr.19: 10106368-10106368 on Build GRCh38
rs779031052 Chr.19: 10106350-10106350 on Build GRCh38
rs1049647300 Chr.19: 10106257-10106257 on Build GRCh38
rs780263009 Chr.19: 10106379-10106379 on Build GRCh38
rs746069216 Chr.19: 10106364-10106364 on Build GRCh38
rs753986549 Chr.19: 10106377-10106377 on Build GRCh38
rs1033480152 Chr.19: 10106276-10106276 on Build GRCh38
rs980589460 Chr.19: 10106394-10106394 on Build GRCh38
rs551378309 Chr.19: 10106250-10106250 on Build GRCh38
rs537856657 Chr.19: 10106313-10106313 on Build GRCh38
rs953829900 Chr.19: 10106361-10106361 on Build GRCh38
rs867284680 Chr.19: 10106273-10106273 on Build GRCh38
rs2290686 Chr.19: 10106301-10106301 on Build GRCh38
rs1028531433 Chr.19: 10106306-10106306 on Build GRCh38
rs74178193 Chr.19: 10106237-10106237 on Build GRCh38
rs141612097 Chr.19: 10106239-10106239 on Build GRCh38
rs761319840 Chr.19: 10106362-10106363 on Build GRCh38
rs544188416 Chr.19: 10106219-10106219 on Build GRCh38
rs199958570 Chr.19: 10106384-10106384 on Build GRCh38
rs1040677808 Chr.19: 10106403-10106403 on Build GRCh38

View Full Product Details