Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 1535280-1535547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PITPNA
Entrez Gene ID
5306
Gene Name
phosphatidylinositol transfer protein alpha
Gene Aliases
HEL-S-36, PI-TPalpha, PITPN, VIB1A
UniGene
Hs.429819
Gene Chromosome Location
Chr.17: 1517989-1562816 on Build GRCh38
Transcripts
NM_006224.3 XM_011523939.1 XM_011523940.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5910945 c.457-5C>T p.? Chr.17: 1535523-1535523 on Build GRCh38
4714551 c.521G>A p.G174D Chr.17: 1535454-1535454 on Build GRCh38
4893506 c.517T>C p.L173L Chr.17: 1535458-1535458 on Build GRCh38
3514463 c.517T>C p.L173L Chr.17: 1535458-1535458 on Build GRCh38
4897793 c.523C>T p.P175S Chr.17: 1535452-1535452 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769757130 Chr.17: 1535312-1535312 on Build GRCh38
rs761123816 Chr.17: 1535477-1535477 on Build GRCh38
rs377695162 Chr.17: 1535315-1535315 on Build GRCh38
rs776640186 Chr.17: 1535379-1535379 on Build GRCh38
rs569988380 Chr.17: 1535348-1535348 on Build GRCh38
rs919972273 Chr.17: 1535353-1535353 on Build GRCh38
rs754499881 Chr.17: 1535419-1535419 on Build GRCh38
rs774499211 Chr.17: 1535452-1535452 on Build GRCh38
rs939914635 Chr.17: 1535344-1535344 on Build GRCh38
rs148261964 Chr.17: 1535479-1535479 on Build GRCh38
rs7504062 Chr.17: 1535438-1535438 on Build GRCh38
rs751075600 Chr.17: 1535417-1535417 on Build GRCh38
rs370619562 Chr.17: 1535458-1535458 on Build GRCh38
rs745938168 Chr.17: 1535454-1535454 on Build GRCh38
rs759128400 Chr.17: 1535416-1535417 on Build GRCh38
rs756150027 Chr.17: 1535428-1535428 on Build GRCh38
rs367634044 Chr.17: 1535450-1535450 on Build GRCh38
rs890294508 Chr.17: 1535361-1535361 on Build GRCh38
rs753252838 Chr.17: 1535392-1535392 on Build GRCh38
rs754225051 Chr.17: 1535413-1535413 on Build GRCh38
rs781248739 Chr.17: 1535421-1535421 on Build GRCh38
rs764403138 Chr.17: 1535403-1535403 on Build GRCh38
rs201928722 Chr.17: 1535448-1535448 on Build GRCh38
rs562941547 Chr.17: 1535331-1535331 on Build GRCh38
rs752423557 Chr.17: 1535449-1535450 on Build GRCh38
rs374746766 Chr.17: 1535400-1535400 on Build GRCh38
rs776912775 Chr.17: 1535501-1535501 on Build GRCh38
rs879532237 Chr.17: 1535422-1535422 on Build GRCh38
rs777655730 Chr.17: 1535430-1535430 on Build GRCh38
rs762147060 Chr.17: 1535508-1535508 on Build GRCh38
rs775579892 Chr.17: 1535459-1535459 on Build GRCh38
rs767241463 Chr.17: 1535427-1535427 on Build GRCh38
rs751098830 Chr.17: 1535329-1535329 on Build GRCh38
rs529561036 Chr.17: 1535414-1535414 on Build GRCh38
rs370352356 Chr.17: 1535319-1535319 on Build GRCh38
rs748132809 Chr.17: 1535423-1535423 on Build GRCh38
rs928550272 Chr.17: 1535333-1535333 on Build GRCh38
rs1048057325 Chr.17: 1535358-1535358 on Build GRCh38
rs759604776 Chr.17: 1535332-1535332 on Build GRCh38
rs755883575 Chr.17: 1535521-1535522 on Build GRCh38
rs768060072 Chr.17: 1535340-1535340 on Build GRCh38
rs915662025 Chr.17: 1535322-1535322 on Build GRCh38
rs773534790 Chr.17: 1535313-1535313 on Build GRCh38

View Full Product Details