Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 139255467-139255740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TXLNB
Entrez Gene ID
167838
Gene Name
taxilin beta
Gene Aliases
C6orf198, LST001, MDP77, dJ522B19.2
UniGene
Hs.535820
Gene Chromosome Location
Chr.6: 139159160-139324011 on Build GRCh38
Transcripts
NM_153235.3 XM_005266836.1 XM_011535505.2 XM_011535506.2 XM_017010322.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7233058 c.1042G>A p.V348M Chr.6: 139255599-139255599 on Build GRCh38
332095 c.1012C>A p.Q338K Chr.6: 139255629-139255629 on Build GRCh38
3621144 c.1033C>T p.Q345* Chr.6: 139255608-139255608 on Build GRCh38
3357956 c.227G>A p.G76D Chr.6: 139255551-139255551 on Build GRCh38
5061452 c.1015G>A p.A339T Chr.6: 139255626-139255626 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375686352 Chr.6: 139255667-139255667 on Build GRCh38
rs771518957 Chr.6: 139255666-139255666 on Build GRCh38
rs762392519 Chr.6: 139255596-139255596 on Build GRCh38
rs777521287 Chr.6: 139255545-139255545 on Build GRCh38
rs773727353 Chr.6: 139255676-139255676 on Build GRCh38
rs368824807 Chr.6: 139255604-139255604 on Build GRCh38
rs745311193 Chr.6: 139255568-139255568 on Build GRCh38
rs376264844 Chr.6: 139255657-139255657 on Build GRCh38
rs200315092 Chr.6: 139255669-139255669 on Build GRCh38
rs748961252 Chr.6: 139255547-139255547 on Build GRCh38
rs143946779 Chr.6: 139255642-139255642 on Build GRCh38
rs776984017 Chr.6: 139255714-139255714 on Build GRCh38
rs200835658 Chr.6: 139255548-139255548 on Build GRCh38
rs755943231 Chr.6: 139255542-139255542 on Build GRCh38
rs1000772012 Chr.6: 139255587-139255587 on Build GRCh38
rs752680537 Chr.6: 139255638-139255638 on Build GRCh38
rs770361252 Chr.6: 139255675-139255675 on Build GRCh38
rs372348432 Chr.6: 139255650-139255650 on Build GRCh38
rs778485475 Chr.6: 139255659-139255659 on Build GRCh38
rs941629196 Chr.6: 139255705-139255705 on Build GRCh38
rs773914168 Chr.6: 139255564-139255564 on Build GRCh38
rs767553918 Chr.6: 139255631-139255631 on Build GRCh38
rs866013283 Chr.6: 139255608-139255608 on Build GRCh38
rs751529743 Chr.6: 139255536-139255536 on Build GRCh38
rs545926135 Chr.6: 139255713-139255713 on Build GRCh38
rs746221578 Chr.6: 139255535-139255536 on Build GRCh38
rs765723224 Chr.6: 139255598-139255598 on Build GRCh38
rs749014562 Chr.6: 139255652-139255652 on Build GRCh38
rs879096475 Chr.6: 139255602-139255602 on Build GRCh38
rs534790924 Chr.6: 139255496-139255496 on Build GRCh38
rs776494154 Chr.6: 139255528-139255529 on Build GRCh38
rs553328654 Chr.6: 139255513-139255513 on Build GRCh38
rs777146501 Chr.6: 139255494-139255494 on Build GRCh38
rs777340764 Chr.6: 139255580-139255580 on Build GRCh38
rs377187778 Chr.6: 139255693-139255693 on Build GRCh38
rs745526942 Chr.6: 139255660-139255660 on Build GRCh38
rs747800100 Chr.6: 139255538-139255538 on Build GRCh38
rs780962830 Chr.6: 139255502-139255502 on Build GRCh38
rs748074347 Chr.6: 139255540-139255540 on Build GRCh38
rs781050366 Chr.6: 139255539-139255539 on Build GRCh38
rs758503589 Chr.6: 139255515-139255515 on Build GRCh38
rs755081196 Chr.6: 139255625-139255625 on Build GRCh38
rs993966318 Chr.6: 139255658-139255658 on Build GRCh38
rs763243962 Chr.6: 139255603-139255603 on Build GRCh38
rs751795356 Chr.6: 139255618-139255618 on Build GRCh38
rs758221033 Chr.6: 139255583-139255583 on Build GRCh38
rs771595096 Chr.6: 139255572-139255572 on Build GRCh38
rs17068451 Chr.6: 139255599-139255599 on Build GRCh38

View Full Product Details