Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 147670492-147670721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRMT9
Entrez Gene ID
90826
Gene Name
protein arginine methyltransferase 9
Gene Aliases
PRMT10
UniGene
Hs.591692
Gene Chromosome Location
Chr.4: 147635626-147684230 on Build GRCh38
Transcripts
NM_001304458.1 NM_138364.3 XM_017008823.1 XM_017008824.1 XM_017008825.1 XM_017008826.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6055535 c.817G>T p.A273S Chr.4: 147670670-147670670 on Build GRCh38
1051857 c.846+2T>C p.? Chr.4: 147670639-147670639 on Build GRCh38
6756262 c.820T>A p.W274R Chr.4: 147670667-147670667 on Build GRCh38
7156993 c.802G>C p.E268Q Chr.4: 147670685-147670685 on Build GRCh38
1051858 c.790G>T p.E264* Chr.4: 147670697-147670697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757874726 Chr.4: 147670668-147670668 on Build GRCh38
rs2036502 Chr.4: 147670618-147670618 on Build GRCh38
rs138129482 Chr.4: 147670534-147670534 on Build GRCh38
rs966801918 Chr.4: 147670538-147670538 on Build GRCh38
rs143662036 Chr.4: 147670547-147670547 on Build GRCh38
rs779886610 Chr.4: 147670680-147670680 on Build GRCh38
rs925101454 Chr.4: 147670552-147670552 on Build GRCh38
rs369345136 Chr.4: 147670526-147670526 on Build GRCh38
rs367691420 Chr.4: 147670601-147670601 on Build GRCh38
rs929382991 Chr.4: 147670558-147670558 on Build GRCh38
rs913920149 Chr.4: 147670532-147670532 on Build GRCh38
rs539568158 Chr.4: 147670529-147670529 on Build GRCh38
rs537074206 Chr.4: 147670517-147670517 on Build GRCh38
rs376324319 Chr.4: 147670653-147670653 on Build GRCh38
rs992143327 Chr.4: 147670527-147670527 on Build GRCh38
rs780261399 Chr.4: 147670651-147670651 on Build GRCh38
rs556747429 Chr.4: 147670525-147670525 on Build GRCh38
rs543782558 Chr.4: 147670631-147670631 on Build GRCh38
rs7435374 Chr.4: 147670608-147670608 on Build GRCh38
rs143458755 Chr.4: 147670652-147670652 on Build GRCh38
rs191821016 Chr.4: 147670629-147670629 on Build GRCh38
rs755728639 Chr.4: 147670622-147670622 on Build GRCh38
rs541711610 Chr.4: 147670602-147670602 on Build GRCh38
rs748002634 Chr.4: 147670607-147670607 on Build GRCh38
rs780179891 Chr.4: 147670685-147670685 on Build GRCh38
rs370775299 Chr.4: 147670660-147670660 on Build GRCh38
rs200390614 Chr.4: 147670681-147670681 on Build GRCh38
rs147164877 Chr.4: 147670674-147670674 on Build GRCh38
rs779603849 Chr.4: 147670671-147670671 on Build GRCh38

View Full Product Details