Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 48604833-48605092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR13
Entrez Gene ID
64743
Gene Name
WD repeat domain 13
Gene Aliases
MG21
UniGene
Hs.521973
Gene Chromosome Location
Chr.X: 48597492-48605194 on Build GRCh38
Transcripts
NM_001166426.1 NM_017883.4 NR_029427.1 XM_006724539.1 XM_006724541.3 XM_011543947.2

SNP Detail

SNP ID Chromosome Location
rs782070385 Chr.X: 48605056-48605056 on Build GRCh38
rs370493659 Chr.X: 48605041-48605041 on Build GRCh38
rs782519758 Chr.X: 48604924-48604924 on Build GRCh38
rs144018865 Chr.X: 48604852-48604852 on Build GRCh38
rs782416576 Chr.X: 48605044-48605044 on Build GRCh38
rs782720352 Chr.X: 48604906-48604906 on Build GRCh38
rs782584055 Chr.X: 48604990-48604990 on Build GRCh38
rs369636679 Chr.X: 48604870-48604870 on Build GRCh38
rs782356868 Chr.X: 48605009-48605009 on Build GRCh38
rs782491786 Chr.X: 48605065-48605065 on Build GRCh38
rs782117938 Chr.X: 48605066-48605066 on Build GRCh38
rs781816285 Chr.X: 48604973-48604973 on Build GRCh38
rs782553788 Chr.X: 48604873-48604873 on Build GRCh38
rs782210313 Chr.X: 48604996-48604996 on Build GRCh38
rs374106808 Chr.X: 48605039-48605039 on Build GRCh38
rs782734233 Chr.X: 48605057-48605057 on Build GRCh38
rs782703286 Chr.X: 48604874-48604874 on Build GRCh38
rs782478121 Chr.X: 48604896-48604896 on Build GRCh38
rs781811852 Chr.X: 48604891-48604891 on Build GRCh38
rs782614484 Chr.X: 48604897-48604897 on Build GRCh38
rs782669008 Chr.X: 48604954-48604954 on Build GRCh38
rs781923689 Chr.X: 48605046-48605046 on Build GRCh38
rs782291105 Chr.X: 48604964-48604964 on Build GRCh38

View Full Product Details