Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 65959113-65959356 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSGA10IP
Entrez Gene ID
254187
Gene Name
testis specific 10 interacting protein
Gene Aliases
FAM161C
UniGene
Hs.350671
Gene Chromosome Location
Chr.11: 65945445-65960437 on Build GRCh38
Transcripts
NM_152762.2 XM_011544887.2 XM_011544888.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5006260 c.1333-4T>C p.? Chr.11: 65959186-65959186 on Build GRCh38
6810387 c.1413T>C p.S471S Chr.11: 65959270-65959270 on Build GRCh38
930587 c.1435G>T p.E479* Chr.11: 65959292-65959292 on Build GRCh38
930586 c.1352T>C p.V451A Chr.11: 65959209-65959209 on Build GRCh38
2166885 c.1439G>A p.G480D Chr.11: 65959296-65959296 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777183851 Chr.11: 65959322-65959322 on Build GRCh38
rs866372928 Chr.11: 65959336-65959336 on Build GRCh38
rs141010009 Chr.11: 65959173-65959173 on Build GRCh38
rs965603951 Chr.11: 65959155-65959155 on Build GRCh38
rs922136338 Chr.11: 65959228-65959228 on Build GRCh38
rs780044330 Chr.11: 65959250-65959250 on Build GRCh38
rs764137373 Chr.11: 65959229-65959229 on Build GRCh38
rs765578835 Chr.11: 65959217-65959217 on Build GRCh38
rs771896320 Chr.11: 65959295-65959295 on Build GRCh38
rs574648093 Chr.11: 65959334-65959334 on Build GRCh38
rs778637522 Chr.11: 65959175-65959175 on Build GRCh38
rs192672036 Chr.11: 65959221-65959221 on Build GRCh38
rs752911707 Chr.11: 65959218-65959218 on Build GRCh38
rs755962821 Chr.11: 65959246-65959246 on Build GRCh38
rs994672970 Chr.11: 65959325-65959325 on Build GRCh38
rs746538610 Chr.11: 65959314-65959314 on Build GRCh38
rs370656960 Chr.11: 65959330-65959330 on Build GRCh38
rs766190402 Chr.11: 65959265-65959268 on Build GRCh38
rs759961519 Chr.11: 65959216-65959216 on Build GRCh38
rs756126008 Chr.11: 65959145-65959145 on Build GRCh38
rs373627822 Chr.11: 65959146-65959146 on Build GRCh38
rs750331263 Chr.11: 65959142-65959142 on Build GRCh38
rs372485688 Chr.11: 65959200-65959200 on Build GRCh38
rs747858372 Chr.11: 65959197-65959197 on Build GRCh38
rs753517722 Chr.11: 65959257-65959257 on Build GRCh38
rs376193585 Chr.11: 65959282-65959282 on Build GRCh38
rs777380389 Chr.11: 65959297-65959297 on Build GRCh38
rs559689560 Chr.11: 65959156-65959156 on Build GRCh38
rs375188190 Chr.11: 65959158-65959158 on Build GRCh38
rs746429500 Chr.11: 65959326-65959326 on Build GRCh38
rs761219143 Chr.11: 65959208-65959208 on Build GRCh38
rs751659923 Chr.11: 65959231-65959231 on Build GRCh38
rs778913900 Chr.11: 65959265-65959265 on Build GRCh38
rs771288241 Chr.11: 65959318-65959318 on Build GRCh38
rs377217355 Chr.11: 65959215-65959215 on Build GRCh38
rs747768525 Chr.11: 65959277-65959277 on Build GRCh38
rs913532982 Chr.11: 65959214-65959214 on Build GRCh38
rs770166413 Chr.11: 65959327-65959327 on Build GRCh38
rs371026018 Chr.11: 65959258-65959258 on Build GRCh38
rs772885840 Chr.11: 65959207-65959207 on Build GRCh38
rs373566789 Chr.11: 65959264-65959264 on Build GRCh38
rs1020110318 Chr.11: 65959153-65959153 on Build GRCh38

View Full Product Details