Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46595463-46595736 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHCBP1
Entrez Gene ID
79801
Gene Name
SHC binding and spindle associated 1
Gene Aliases
PAL
UniGene
Hs.123253
Gene Chromosome Location
Chr.16: 46578591-46624933 on Build GRCh38
Transcripts
NM_001324318.1 NM_001324319.1 NM_024745.4 NR_136738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5384890 c.1427A>G p.E476G Chr.16: 46595589-46595589 on Build GRCh38
6777761 c.1459G>A p.A487T Chr.16: 46595557-46595557 on Build GRCh38
5182734 c.1397C>T p.T466M Chr.16: 46595619-46595619 on Build GRCh38
7389749 c.1352A>T p.H451L Chr.16: 46595664-46595664 on Build GRCh38
342209 c.1459G>T p.A487S Chr.16: 46595557-46595557 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778481033 Chr.16: 46595611-46595611 on Build GRCh38
rs567938048 Chr.16: 46595591-46595591 on Build GRCh38
rs772882846 Chr.16: 46595505-46595505 on Build GRCh38
rs866603453 Chr.16: 46595688-46595688 on Build GRCh38
rs768840759 Chr.16: 46595689-46595692 on Build GRCh38
rs758981763 Chr.16: 46595516-46595516 on Build GRCh38
rs112829840 Chr.16: 46595540-46595540 on Build GRCh38
rs767463430 Chr.16: 46595676-46595676 on Build GRCh38
rs750726620 Chr.16: 46595536-46595536 on Build GRCh38
rs777521264 Chr.16: 46595587-46595587 on Build GRCh38
rs918938756 Chr.16: 46595694-46595694 on Build GRCh38
rs764676760 Chr.16: 46595520-46595520 on Build GRCh38
rs372725241 Chr.16: 46595704-46595704 on Build GRCh38
rs755976652 Chr.16: 46595570-46595570 on Build GRCh38
rs747993306 Chr.16: 46595562-46595562 on Build GRCh38
rs145985657 Chr.16: 46595615-46595615 on Build GRCh38
rs756001686 Chr.16: 46595685-46595685 on Build GRCh38
rs748908174 Chr.16: 46595507-46595507 on Build GRCh38
rs1009622540 Chr.16: 46595574-46595574 on Build GRCh38
rs770486553 Chr.16: 46595508-46595508 on Build GRCh38
rs1022397782 Chr.16: 46595606-46595606 on Build GRCh38
rs533467936 Chr.16: 46595699-46595699 on Build GRCh38
rs759691750 Chr.16: 46595668-46595668 on Build GRCh38
rs553507161 Chr.16: 46595710-46595710 on Build GRCh38
rs777621387 Chr.16: 46595687-46595687 on Build GRCh38
rs1026863027 Chr.16: 46595689-46595689 on Build GRCh38
rs1042319783 Chr.16: 46595594-46595594 on Build GRCh38
rs778570927 Chr.16: 46595708-46595708 on Build GRCh38
rs745407076 Chr.16: 46595614-46595614 on Build GRCh38
rs771666579 Chr.16: 46595711-46595711 on Build GRCh38
rs766629175 Chr.16: 46595539-46595539 on Build GRCh38
rs199534171 Chr.16: 46595499-46595499 on Build GRCh38
rs762272396 Chr.16: 46595529-46595529 on Build GRCh38
rs765492713 Chr.16: 46595533-46595533 on Build GRCh38
rs202030254 Chr.16: 46595644-46595644 on Build GRCh38
rs926930869 Chr.16: 46595650-46595650 on Build GRCh38
rs751692076 Chr.16: 46595554-46595554 on Build GRCh38
rs751782063 Chr.16: 46595662-46595662 on Build GRCh38
rs758677480 Chr.16: 46595537-46595537 on Build GRCh38
rs753649292 Chr.16: 46595691-46595691 on Build GRCh38
rs267604549 Chr.16: 46595663-46595663 on Build GRCh38
rs773847251 Chr.16: 46595512-46595512 on Build GRCh38
rs777237271 Chr.16: 46595618-46595618 on Build GRCh38
rs762208281 Chr.16: 46595637-46595637 on Build GRCh38
rs756162190 Chr.16: 46595557-46595557 on Build GRCh38
rs763340134 Chr.16: 46595655-46595655 on Build GRCh38
rs781210656 Chr.16: 46595558-46595558 on Build GRCh38
rs770254582 Chr.16: 46595647-46595647 on Build GRCh38
rs369921811 Chr.16: 46595577-46595577 on Build GRCh38
rs752729001 Chr.16: 46595677-46595677 on Build GRCh38
rs777914584 Chr.16: 46595571-46595571 on Build GRCh38
rs773704460 Chr.16: 46595649-46595649 on Build GRCh38
rs535052871 Chr.16: 46595601-46595601 on Build GRCh38
rs372325367 Chr.16: 46595576-46595576 on Build GRCh38
rs368712899 Chr.16: 46595521-46595521 on Build GRCh38
rs766719161 Chr.16: 46595661-46595661 on Build GRCh38
rs778225531 Chr.16: 46595514-46595514 on Build GRCh38

View Full Product Details