Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50358655-50358906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYB561D2
Entrez Gene ID
11068
Gene Name
cytochrome b561 family member D2
Gene Aliases
101F6, TSP10, XXcos-LUCA11.4
UniGene
Hs.149443
Gene Chromosome Location
Chr.3: 50350695-50368197 on Build GRCh38
Transcripts
NR_111912.1 NM_007024.4

Gene Symbol
TMEM115
Entrez Gene ID
11070
Gene Name
transmembrane protein 115
Gene Aliases
PL6
UniGene
Hs.91566
Gene Chromosome Location
Chr.3: 50354749-50359508 on Build GRCh38
Transcripts
NR_111912.1 NM_007024.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6842894 c.178T>G p.W60G Chr.3: 50358886-50358886 on Build GRCh38
6803124 c.323C>T p.S108F Chr.3: 50358741-50358741 on Build GRCh38
3993177 c.277T>C p.W93R Chr.3: 50358787-50358787 on Build GRCh38
6803123 c.298A>G p.I100V Chr.3: 50358766-50358766 on Build GRCh38
247861 c.280G>A p.G94R Chr.3: 50358784-50358784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781587387 Chr.3: 50358851-50358851 on Build GRCh38
rs780411130 Chr.3: 50358716-50358716 on Build GRCh38
rs770689015 Chr.3: 50358788-50358788 on Build GRCh38
rs752169433 Chr.3: 50358692-50358692 on Build GRCh38
rs1052040130 Chr.3: 50358781-50358781 on Build GRCh38
rs751764992 Chr.3: 50358761-50358761 on Build GRCh38
rs767079372 Chr.3: 50358687-50358687 on Build GRCh38
rs759173769 Chr.3: 50358686-50358686 on Build GRCh38
rs774126700 Chr.3: 50358807-50358807 on Build GRCh38
rs756161331 Chr.3: 50358875-50358875 on Build GRCh38
rs587738327 Chr.3: 50358780-50358780 on Build GRCh38
rs748218306 Chr.3: 50358773-50358773 on Build GRCh38
rs781462803 Chr.3: 50358766-50358766 on Build GRCh38
rs750910063 Chr.3: 50358831-50358831 on Build GRCh38
rs896265830 Chr.3: 50358810-50358810 on Build GRCh38
rs763596493 Chr.3: 50358819-50358819 on Build GRCh38
rs765681327 Chr.3: 50358700-50358700 on Build GRCh38
rs150309447 Chr.3: 50358745-50358745 on Build GRCh38
rs917793740 Chr.3: 50358885-50358885 on Build GRCh38
rs762270049 Chr.3: 50358757-50358759 on Build GRCh38
rs755249259 Chr.3: 50358733-50358733 on Build GRCh38
rs757806916 Chr.3: 50358696-50358696 on Build GRCh38
rs771686400 Chr.3: 50358814-50358814 on Build GRCh38
rs199936568 Chr.3: 50358809-50358809 on Build GRCh38
rs376314064 Chr.3: 50358844-50358844 on Build GRCh38
rs763506366 Chr.3: 50358833-50358833 on Build GRCh38
rs587669973 Chr.3: 50358860-50358860 on Build GRCh38
rs587663416 Chr.3: 50358775-50358775 on Build GRCh38
rs759223068 Chr.3: 50358813-50358813 on Build GRCh38
rs766129741 Chr.3: 50358678-50358678 on Build GRCh38
rs934986398 Chr.3: 50358779-50358779 on Build GRCh38
rs773887068 Chr.3: 50358685-50358685 on Build GRCh38
rs750907228 Chr.3: 50358710-50358710 on Build GRCh38
rs751891845 Chr.3: 50358722-50358722 on Build GRCh38
rs868361120 Chr.3: 50358719-50358719 on Build GRCh38
rs587619729 Chr.3: 50358840-50358840 on Build GRCh38
rs762687791 Chr.3: 50358677-50358677 on Build GRCh38
rs777893446 Chr.3: 50358881-50358881 on Build GRCh38
rs373868314 Chr.3: 50358818-50358818 on Build GRCh38
rs775070490 Chr.3: 50358815-50358815 on Build GRCh38
rs992046127 Chr.3: 50358782-50358782 on Build GRCh38
rs866571195 Chr.3: 50358800-50358800 on Build GRCh38
rs369982914 Chr.3: 50358714-50358714 on Build GRCh38
rs751943104 Chr.3: 50358842-50358842 on Build GRCh38

View Full Product Details