Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 24994142-24994409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C12orf77
Entrez Gene ID
196415
Gene Name
chromosome 12 open reading frame 77
Gene Aliases
-
UniGene
Hs.434453
Gene Chromosome Location
Chr.12: 24993424-24997673 on Build GRCh38
Transcripts
NM_001101339.2 XM_017018966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
692707 c.420T>A p.A140A Chr.12: 24994315-24994315 on Build GRCh38
4388579 c.415C>T p.H139Y Chr.12: 24994320-24994320 on Build GRCh38
7259871 c.408-8delT p.? Chr.12: 24994335-24994335 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756705649 Chr.12: 24994284-24994284 on Build GRCh38
rs766697639 Chr.12: 24994322-24994322 on Build GRCh38
rs768823227 Chr.12: 24994369-24994369 on Build GRCh38
rs528324211 Chr.12: 24994333-24994333 on Build GRCh38
rs980037929 Chr.12: 24994233-24994233 on Build GRCh38
rs779695560 Chr.12: 24994345-24994346 on Build GRCh38
rs749094913 Chr.12: 24994367-24994367 on Build GRCh38
rs750390645 Chr.12: 24994355-24994355 on Build GRCh38
rs758039659 Chr.12: 24994257-24994257 on Build GRCh38
rs935352417 Chr.12: 24994240-24994240 on Build GRCh38
rs1003875178 Chr.12: 24994379-24994379 on Build GRCh38
rs763801168 Chr.12: 24994340-24994340 on Build GRCh38
rs751146627 Chr.12: 24994348-24994348 on Build GRCh38
rs772208687 Chr.12: 24994312-24994312 on Build GRCh38
rs773866284 Chr.12: 24994314-24994314 on Build GRCh38
rs768900415 Chr.12: 24994287-24994287 on Build GRCh38
rs757252946 Chr.12: 24994273-24994273 on Build GRCh38
rs766704662 Chr.12: 24994251-24994251 on Build GRCh38
rs699030 Chr.12: 24994366-24994366 on Build GRCh38
rs745750682 Chr.12: 24994282-24994282 on Build GRCh38
rs866038910 Chr.12: 24994388-24994388 on Build GRCh38
rs762575419 Chr.12: 24994330-24994330 on Build GRCh38
rs756127660 Chr.12: 24994386-24994386 on Build GRCh38
rs16928315 Chr.12: 24994258-24994258 on Build GRCh38
rs748320662 Chr.12: 24994377-24994377 on Build GRCh38
rs531395908 Chr.12: 24994357-24994357 on Build GRCh38
rs761977442 Chr.12: 24994295-24994295 on Build GRCh38
rs774630493 Chr.12: 24994289-24994289 on Build GRCh38
rs560797320 Chr.12: 24994354-24994354 on Build GRCh38
rs114597208 Chr.12: 24994327-24994327 on Build GRCh38
rs185499424 Chr.12: 24994231-24994231 on Build GRCh38
rs377073729 Chr.12: 24994278-24994278 on Build GRCh38
rs746512682 Chr.12: 24994262-24994262 on Build GRCh38
rs190361414 Chr.12: 24994254-24994254 on Build GRCh38
rs761211403 Chr.12: 24994316-24994316 on Build GRCh38
rs779277716 Chr.12: 24994371-24994371 on Build GRCh38
rs749510886 Chr.12: 24994285-24994285 on Build GRCh38

View Full Product Details