Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 71761473-71761728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf54
Entrez Gene ID
64115
Gene Name
chromosome 10 open reading frame 54
Gene Aliases
B7-H5, B7H5, DD1alpha, GI24, PD-1H, PP2135, SISP1, VISTA
UniGene
Hs.47382
Gene Chromosome Location
Chr.10: 71747556-71773580 on Build GRCh38
Transcripts
NM_022153.1 NM_022124.5 XM_006717940.3 XM_006717942.3 XM_011540039.2 XM_011540042.2 XM_011540043.2 XM_011540044.2 XM_011540045.2 XM_011540046.2 XM_011540047.2 XM_011540048.2 XM_011540049.2 XM_011540051.2 XM_011540052.2 XM_017016499.1 XM_017016500.1 XM_017016501.1 XM_017016502.1 XM_017016503.1 XM_017016504.1 XM_017016505.1

Gene Symbol
CDH23
Entrez Gene ID
64072
Gene Name
cadherin-related 23
Gene Aliases
CDHR23, USH1D
UniGene
Hs.656032
Gene Chromosome Location
Chr.10: 71396934-71815947 on Build GRCh38
Transcripts
NM_022153.1 NM_022124.5 XM_006717940.3 XM_006717942.3 XM_011540039.2 XM_011540042.2 XM_011540043.2 XM_011540044.2 XM_011540045.2 XM_011540046.2 XM_011540047.2 XM_011540048.2 XM_011540049.2 XM_011540051.2 XM_011540052.2 XM_017016499.1 XM_017016500.1 XM_017016501.1 XM_017016502.1 XM_017016503.1 XM_017016504.1 XM_017016505.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6271285 c.432C>A p.Y144* Chr.10: 71761677-71761677 on Build GRCh38
5998921 c.400C>T p.R134C Chr.10: 71761709-71761709 on Build GRCh38
4975283 c.468G>A p.S156S Chr.10: 71761641-71761641 on Build GRCh38
4493265 c.411C>T p.T137T Chr.10: 71761698-71761698 on Build GRCh38
6407767 c.412C>A p.L138M Chr.10: 71761697-71761697 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757580511 Chr.10: 71761708-71761708 on Build GRCh38
rs41281324 Chr.10: 71761581-71761581 on Build GRCh38
rs982727677 Chr.10: 71761583-71761583 on Build GRCh38
rs368289633 Chr.10: 71761567-71761567 on Build GRCh38
rs35478708 Chr.10: 71761660-71761661 on Build GRCh38
rs751705005 Chr.10: 71761696-71761696 on Build GRCh38
rs200235042 Chr.10: 71761708-71761708 on Build GRCh38
rs1034532970 Chr.10: 71761509-71761509 on Build GRCh38
rs777637150 Chr.10: 71761607-71761607 on Build GRCh38
rs147441567 Chr.10: 71761642-71761642 on Build GRCh38
rs769995499 Chr.10: 71761665-71761665 on Build GRCh38
rs757167523 Chr.10: 71761619-71761619 on Build GRCh38
rs755473957 Chr.10: 71761568-71761568 on Build GRCh38
rs142695132 Chr.10: 71761615-71761615 on Build GRCh38
rs763373080 Chr.10: 71761556-71761556 on Build GRCh38
rs1026782030 Chr.10: 71761659-71761661 on Build GRCh38
rs766836355 Chr.10: 71761563-71761563 on Build GRCh38
rs139256346 Chr.10: 71761519-71761520 on Build GRCh38
rs3747867 Chr.10: 71761614-71761614 on Build GRCh38
rs773460839 Chr.10: 71761686-71761686 on Build GRCh38
rs369497696 Chr.10: 71761692-71761692 on Build GRCh38
rs773548979 Chr.10: 71761552-71761552 on Build GRCh38
rs1014526824 Chr.10: 71761574-71761574 on Build GRCh38
rs766990416 Chr.10: 71761577-71761577 on Build GRCh38
rs376085614 Chr.10: 71761629-71761629 on Build GRCh38
rs781273917 Chr.10: 71761647-71761647 on Build GRCh38
rs530354809 Chr.10: 71761687-71761687 on Build GRCh38
rs778595953 Chr.10: 71761626-71761626 on Build GRCh38
rs771907605 Chr.10: 71761641-71761641 on Build GRCh38
rs771290690 Chr.10: 71761694-71761694 on Build GRCh38
rs748283137 Chr.10: 71761656-71761656 on Build GRCh38
rs546854374 Chr.10: 71761580-71761580 on Build GRCh38

View Full Product Details