Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57764329-57764602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP27B1
Entrez Gene ID
1594
Gene Name
cytochrome P450 family 27 subfamily B member 1
Gene Aliases
CP2B, CYP1, CYP1alpha, CYP27B, P450c1, PDDR, VDD1, VDDR, VDDRI, VDR
UniGene
Hs.524528
Gene Chromosome Location
Chr.12: 57762334-57767193 on Build GRCh38
Transcripts
NM_000785.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1363435 c.1046C>G p.A349G Chr.12: 57764468-57764468 on Build GRCh38
1989864 c.1057C>G p.P353A Chr.12: 57764457-57764457 on Build GRCh38
6846469 c.1085C>A p.T362N Chr.12: 57764429-57764429 on Build GRCh38
5882307 c.1103C>T p.P368L Chr.12: 57764411-57764411 on Build GRCh38
468724 c.1038G>A p.E346E Chr.12: 57764476-57764476 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769591385 Chr.12: 57764420-57764420 on Build GRCh38
rs770746377 Chr.12: 57764436-57764436 on Build GRCh38
rs752175742 Chr.12: 57764563-57764563 on Build GRCh38
rs777588673 Chr.12: 57764502-57764502 on Build GRCh38
rs749181794 Chr.12: 57764582-57764583 on Build GRCh38
rs780857122 Chr.12: 57764410-57764410 on Build GRCh38
rs749017261 Chr.12: 57764503-57764503 on Build GRCh38
rs762118198 Chr.12: 57764540-57764540 on Build GRCh38
rs775708027 Chr.12: 57764524-57764524 on Build GRCh38
rs141927200 Chr.12: 57764504-57764504 on Build GRCh38
rs578010655 Chr.12: 57764494-57764494 on Build GRCh38
rs908451945 Chr.12: 57764360-57764360 on Build GRCh38
rs767085633 Chr.12: 57764561-57764561 on Build GRCh38
rs867550876 Chr.12: 57764363-57764363 on Build GRCh38
rs533410734 Chr.12: 57764359-57764359 on Build GRCh38
rs567195245 Chr.12: 57764455-57764455 on Build GRCh38
rs776942384 Chr.12: 57764529-57764529 on Build GRCh38
rs1001021197 Chr.12: 57764481-57764481 on Build GRCh38
rs752977580 Chr.12: 57764446-57764446 on Build GRCh38
rs201819682 Chr.12: 57764505-57764505 on Build GRCh38
rs111503667 Chr.12: 57764575-57764575 on Build GRCh38
rs556530774 Chr.12: 57764474-57764474 on Build GRCh38
rs145790221 Chr.12: 57764517-57764517 on Build GRCh38
rs372527928 Chr.12: 57764479-57764479 on Build GRCh38
rs750908103 Chr.12: 57764547-57764547 on Build GRCh38
rs151335249 Chr.12: 57764457-57764457 on Build GRCh38
rs745757851 Chr.12: 57764511-57764511 on Build GRCh38
rs778225054 Chr.12: 57764371-57764371 on Build GRCh38
rs765572208 Chr.12: 57764543-57764543 on Build GRCh38
rs13377933 Chr.12: 57764447-57764447 on Build GRCh38
rs753298199 Chr.12: 57764406-57764406 on Build GRCh38
rs977891042 Chr.12: 57764361-57764361 on Build GRCh38
rs752926249 Chr.12: 57764357-57764357 on Build GRCh38
rs758931129 Chr.12: 57764552-57764552 on Build GRCh38
rs1033347792 Chr.12: 57764580-57764580 on Build GRCh38
rs754449343 Chr.12: 57764385-57764385 on Build GRCh38
rs755816133 Chr.12: 57764573-57764573 on Build GRCh38
rs780623929 Chr.12: 57764470-57764470 on Build GRCh38
rs199575147 Chr.12: 57764398-57764398 on Build GRCh38
rs764387303 Chr.12: 57764527-57764527 on Build GRCh38
rs2229103 Chr.12: 57764393-57764393 on Build GRCh38
rs772982189 Chr.12: 57764427-57764427 on Build GRCh38
rs537825066 Chr.12: 57764462-57764462 on Build GRCh38
rs373672754 Chr.12: 57764428-57764428 on Build GRCh38
rs118204011 Chr.12: 57764487-57764487 on Build GRCh38
rs142446425 Chr.12: 57764444-57764444 on Build GRCh38
rs772097552 Chr.12: 57764515-57764515 on Build GRCh38
rs148336763 Chr.12: 57764395-57764395 on Build GRCh38
rs762823552 Chr.12: 57764471-57764471 on Build GRCh38
rs933909057 Chr.12: 57764440-57764440 on Build GRCh38
rs747732728 Chr.12: 57764415-57764415 on Build GRCh38
rs779651723 Chr.12: 57764394-57764394 on Build GRCh38
rs759563696 Chr.12: 57764443-57764443 on Build GRCh38
rs28934606 Chr.12: 57764510-57764510 on Build GRCh38
rs774541604 Chr.12: 57764441-57764441 on Build GRCh38
rs191727255 Chr.12: 57764449-57764449 on Build GRCh38
rs777427466 Chr.12: 57764357-57764357 on Build GRCh38
rs996571905 Chr.12: 57764439-57764439 on Build GRCh38
rs754452515 Chr.12: 57764465-57764465 on Build GRCh38
rs777428292 Chr.12: 57764577-57764577 on Build GRCh38
rs750331984 Chr.12: 57764369-57764369 on Build GRCh38

View Full Product Details