Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 413050-413275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf97
Entrez Gene ID
400566
Gene Name
chromosome 17 open reading frame 97
Gene Aliases
CK20, LIAT1
UniGene
Hs.499607
Gene Chromosome Location
Chr.17: 410327-414666 on Build GRCh38
Transcripts
NM_001013672.4 NR_136407.1

Gene Symbol
LOC105371430
Entrez Gene ID
105371430
Gene Name
uncharacterized LOC105371430
Gene Aliases
-
UniGene
Hs.616427
Gene Chromosome Location
Chr.17: 404535-414023 on Build GRCh38
Transcripts
NM_001013672.4 NR_136407.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5981074 c.352T>C p.F118L Chr.17: 413195-413195 on Build GRCh38
6479052 c.383G>A p.R128H Chr.17: 413226-413226 on Build GRCh38
1736037 c.411C>A p.S137R Chr.17: 413254-413254 on Build GRCh38
6487170 c.368G>C p.S123T Chr.17: 413211-413211 on Build GRCh38
3515293 c.388C>T p.P130S Chr.17: 413231-413231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782786322 Chr.17: 413182-413182 on Build GRCh38
rs1043736856 Chr.17: 413225-413225 on Build GRCh38
rs781853712 Chr.17: 413226-413226 on Build GRCh38
rs113767375 Chr.17: 413108-413108 on Build GRCh38
rs782631983 Chr.17: 413154-413154 on Build GRCh38
rs781905970 Chr.17: 413126-413126 on Build GRCh38
rs368996060 Chr.17: 413124-413124 on Build GRCh38
rs782398745 Chr.17: 413162-413162 on Build GRCh38
rs782518002 Chr.17: 413117-413117 on Build GRCh38
rs781881135 Chr.17: 413095-413095 on Build GRCh38
rs782694164 Chr.17: 413195-413195 on Build GRCh38
rs1025917638 Chr.17: 413089-413089 on Build GRCh38
rs782566021 Chr.17: 413094-413094 on Build GRCh38
rs138947672 Chr.17: 413091-413091 on Build GRCh38
rs782041373 Chr.17: 413166-413166 on Build GRCh38
rs914003816 Chr.17: 413223-413223 on Build GRCh38
rs782022380 Chr.17: 413184-413184 on Build GRCh38
rs149869019 Chr.17: 413237-413237 on Build GRCh38
rs782798615 Chr.17: 413125-413125 on Build GRCh38
rs974568523 Chr.17: 413137-413137 on Build GRCh38
rs555806110 Chr.17: 413144-413144 on Build GRCh38
rs782473210 Chr.17: 413090-413090 on Build GRCh38
rs140299852 Chr.17: 413224-413224 on Build GRCh38
rs576500402 Chr.17: 413109-413109 on Build GRCh38
rs782165283 Chr.17: 413192-413192 on Build GRCh38
rs200671415 Chr.17: 413239-413239 on Build GRCh38
rs781941600 Chr.17: 413180-413180 on Build GRCh38
rs782256738 Chr.17: 413160-413160 on Build GRCh38
rs782452138 Chr.17: 413139-413139 on Build GRCh38
rs371581589 Chr.17: 413147-413147 on Build GRCh38
rs541559380 Chr.17: 413233-413233 on Build GRCh38
rs368141276 Chr.17: 413119-413119 on Build GRCh38
rs372647667 Chr.17: 413167-413167 on Build GRCh38
rs781806859 Chr.17: 413213-413213 on Build GRCh38
rs80009610 Chr.17: 413140-413140 on Build GRCh38
rs782028789 Chr.17: 413164-413164 on Build GRCh38
rs782318690 Chr.17: 413170-413170 on Build GRCh38
rs782519076 Chr.17: 413232-413232 on Build GRCh38
rs782124924 Chr.17: 413181-413181 on Build GRCh38

View Full Product Details