Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35767685-35767926 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PROSER3
Entrez Gene ID
148137
Gene Name
proline and serine rich 3
Gene Aliases
C19orf55
UniGene
Hs.527982
Gene Chromosome Location
Chr.19: 35758139-35770115 on Build GRCh38
Transcripts
NM_001039887.2 XM_011526528.1 XM_011526529.2 XM_011526530.1 XM_011526531.1 XM_011526532.1 XM_011526533.2 XM_011526534.1 XM_011526535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7248270 c.974C>T p.A325V Chr.19: 35767820-35767820 on Build GRCh38
3938031 c.1048C>A p.Q350K Chr.19: 35767894-35767894 on Build GRCh38
459982 c.1005G>A p.V335V Chr.19: 35767851-35767851 on Build GRCh38
3388890 c.986C>T p.P329L Chr.19: 35767832-35767832 on Build GRCh38
1392963 c.991G>A p.A331T Chr.19: 35767837-35767837 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs938534434 Chr.19: 35767853-35767853 on Build GRCh38
rs369033179 Chr.19: 35767775-35767775 on Build GRCh38
rs756880215 Chr.19: 35767758-35767761 on Build GRCh38
rs770583246 Chr.19: 35767864-35767864 on Build GRCh38
rs780180435 Chr.19: 35767855-35767855 on Build GRCh38
rs778934458 Chr.19: 35767856-35767856 on Build GRCh38
rs774187444 Chr.19: 35767812-35767812 on Build GRCh38
rs768065219 Chr.19: 35767903-35767903 on Build GRCh38
rs150574903 Chr.19: 35767732-35767732 on Build GRCh38
rs73592448 Chr.19: 35767820-35767820 on Build GRCh38
rs745755657 Chr.19: 35767821-35767822 on Build GRCh38
rs751989418 Chr.19: 35767886-35767886 on Build GRCh38
rs987482285 Chr.19: 35767871-35767871 on Build GRCh38
rs762299399 Chr.19: 35767896-35767896 on Build GRCh38
rs924214457 Chr.19: 35767736-35767736 on Build GRCh38
rs954998845 Chr.19: 35767869-35767869 on Build GRCh38
rs561495817 Chr.19: 35767844-35767844 on Build GRCh38
rs764373767 Chr.19: 35767883-35767883 on Build GRCh38
rs772933424 Chr.19: 35767831-35767831 on Build GRCh38
rs768609828 Chr.19: 35767807-35767807 on Build GRCh38
rs79020519 Chr.19: 35767796-35767796 on Build GRCh38
rs776176044 Chr.19: 35767870-35767870 on Build GRCh38
rs904135039 Chr.19: 35767702-35767702 on Build GRCh38
rs753080471 Chr.19: 35767834-35767834 on Build GRCh38
rs376039474 Chr.19: 35767763-35767763 on Build GRCh38
rs757354855 Chr.19: 35767769-35767769 on Build GRCh38
rs928442931 Chr.19: 35767847-35767847 on Build GRCh38
rs562709481 Chr.19: 35767737-35767737 on Build GRCh38
rs1054048763 Chr.19: 35767745-35767745 on Build GRCh38
rs951290758 Chr.19: 35767826-35767826 on Build GRCh38
rs767149295 Chr.19: 35767827-35767827 on Build GRCh38
rs544127325 Chr.19: 35767734-35767734 on Build GRCh38
rs186555510 Chr.19: 35767743-35767743 on Build GRCh38
rs532017139 Chr.19: 35767858-35767858 on Build GRCh38
rs923904872 Chr.19: 35767877-35767877 on Build GRCh38
rs778190967 Chr.19: 35767755-35767755 on Build GRCh38
rs749087692 Chr.19: 35767804-35767804 on Build GRCh38
rs772834828 Chr.19: 35767863-35767863 on Build GRCh38
rs866522980 Chr.19: 35767860-35767864 on Build GRCh38
rs752077583 Chr.19: 35767837-35767837 on Build GRCh38
rs922025271 Chr.19: 35767709-35767709 on Build GRCh38
rs373406361 Chr.19: 35767878-35767878 on Build GRCh38
rs756338782 Chr.19: 35767854-35767854 on Build GRCh38
rs760397364 Chr.19: 35767833-35767833 on Build GRCh38
rs781248272 Chr.19: 35767842-35767842 on Build GRCh38
rs369977417 Chr.19: 35767823-35767823 on Build GRCh38
rs781248435 Chr.19: 35767798-35767818 on Build GRCh38
rs756179718 Chr.19: 35767905-35767905 on Build GRCh38
rs750471947 Chr.19: 35767904-35767904 on Build GRCh38
rs373157393 Chr.19: 35767801-35767801 on Build GRCh38
rs560638856 Chr.19: 35767787-35767787 on Build GRCh38

View Full Product Details